Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52451641-52451908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT6B
Entrez Gene ID
3854
Gene Name
keratin 6B
Gene Aliases
CK-6B, CK6B, K6B, KRTL1, PC2, PC4
UniGene
Hs.708950
Gene Chromosome Location
Chr.12: 52446651-52452126 on Build GRCh38
Transcripts
NM_005555.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2010599 c.397G>A p.G133R Chr.12: 52451682-52451682 on Build GRCh38
6402947 c.344G>T p.G115V Chr.12: 52451735-52451735 on Build GRCh38
391309 c.363delC p.F122fs*24 Chr.12: 52451716-52451716 on Build GRCh38
1322431 c.412G>T p.V138F Chr.12: 52451667-52451667 on Build GRCh38
694204 c.409G>C p.E137Q Chr.12: 52451670-52451670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369674246 Chr.12: 52451694-52451694 on Build GRCh38
rs560354778 Chr.12: 52451840-52451840 on Build GRCh38
rs779219726 Chr.12: 52451686-52451686 on Build GRCh38
rs774088015 Chr.12: 52451729-52451729 on Build GRCh38
rs780990421 Chr.12: 52451786-52451786 on Build GRCh38
rs540159600 Chr.12: 52451812-52451812 on Build GRCh38
rs764630392 Chr.12: 52451865-52451865 on Build GRCh38
rs778306714 Chr.12: 52451772-52451772 on Build GRCh38
rs61914500 Chr.12: 52451817-52451817 on Build GRCh38
rs562648897 Chr.12: 52451882-52451882 on Build GRCh38
rs766529656 Chr.12: 52451736-52451736 on Build GRCh38
rs756978054 Chr.12: 52451778-52451778 on Build GRCh38
rs934335469 Chr.12: 52451700-52451700 on Build GRCh38
rs146454705 Chr.12: 52451702-52451702 on Build GRCh38
rs753917253 Chr.12: 52451738-52451738 on Build GRCh38
rs377681409 Chr.12: 52451685-52451685 on Build GRCh38
rs61745883 Chr.12: 52451747-52451747 on Build GRCh38
rs774143030 Chr.12: 52451828-52451828 on Build GRCh38
rs988699834 Chr.12: 52451779-52451779 on Build GRCh38
rs768587301 Chr.12: 52451818-52451818 on Build GRCh38
rs914504608 Chr.12: 52451873-52451873 on Build GRCh38
rs554558110 Chr.12: 52451716-52451716 on Build GRCh38
rs546022490 Chr.12: 52451871-52451871 on Build GRCh38
rs766474462 Chr.12: 52451666-52451666 on Build GRCh38
rs780863866 Chr.12: 52451697-52451697 on Build GRCh38
rs771207924 Chr.12: 52451836-52451836 on Build GRCh38
rs753172591 Chr.12: 52451687-52451687 on Build GRCh38
rs574747070 Chr.12: 52451735-52451735 on Build GRCh38
rs756223113 Chr.12: 52451663-52451663 on Build GRCh38
rs769839426 Chr.12: 52451699-52451699 on Build GRCh38
rs753114842 Chr.12: 52451761-52451761 on Build GRCh38
rs752039509 Chr.12: 52451869-52451869 on Build GRCh38
rs144860693 Chr.12: 52451790-52451790 on Build GRCh38
rs772129177 Chr.12: 52451684-52451684 on Build GRCh38
rs777522365 Chr.12: 52451692-52451692 on Build GRCh38
rs374587684 Chr.12: 52451695-52451695 on Build GRCh38
rs765264911 Chr.12: 52451842-52451842 on Build GRCh38
rs769168555 Chr.12: 52451793-52451793 on Build GRCh38
rs780076492 Chr.12: 52451794-52451794 on Build GRCh38
rs947171612 Chr.12: 52451881-52451882 on Build GRCh38
rs761528539 Chr.12: 52451835-52451835 on Build GRCh38
rs71455206 Chr.12: 52451662-52451662 on Build GRCh38
rs753862175 Chr.12: 52451678-52451678 on Build GRCh38
rs758676534 Chr.12: 52451688-52451688 on Build GRCh38
rs752774038 Chr.12: 52451846-52451846 on Build GRCh38
rs765830157 Chr.12: 52451760-52451760 on Build GRCh38
rs202222264 Chr.12: 52451706-52451706 on Build GRCh38
rs374536069 Chr.12: 52451864-52451864 on Build GRCh38
rs758899246 Chr.12: 52451764-52451764 on Build GRCh38
rs745376618 Chr.12: 52451671-52451671 on Build GRCh38
rs776820315 Chr.12: 52451837-52451837 on Build GRCh38
rs779449161 Chr.12: 52451885-52451885 on Build GRCh38
rs141114189 Chr.12: 52451881-52451881 on Build GRCh38

View Full Product Details