Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53434228-53434501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B10
Entrez Gene ID
3028
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Gene Aliases
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1
UniGene
Hs.171280
Gene Chromosome Location
Chr.X: 53431258-53434376 on Build GRCh38
Transcripts
NM_001037811.2 NM_004493.2

SNP Detail

SNP ID Chromosome Location
rs911505039 Chr.X: 53434302-53434302 on Build GRCh38
rs781935025 Chr.X: 53434333-53434333 on Build GRCh38
rs374902916 Chr.X: 53434272-53434272 on Build GRCh38
rs895041688 Chr.X: 53434458-53434458 on Build GRCh38
rs781839633 Chr.X: 53434335-53434335 on Build GRCh38
rs934018976 Chr.X: 53434294-53434294 on Build GRCh38
rs782699513 Chr.X: 53434324-53434324 on Build GRCh38
rs370106783 Chr.X: 53434293-53434293 on Build GRCh38
rs782755855 Chr.X: 53434286-53434286 on Build GRCh38
rs1264014 Chr.X: 53434412-53434412 on Build GRCh38
rs782077284 Chr.X: 53434277-53434277 on Build GRCh38
rs941613640 Chr.X: 53434312-53434312 on Build GRCh38
rs372768875 Chr.X: 53434300-53434300 on Build GRCh38
rs1047977240 Chr.X: 53434406-53434406 on Build GRCh38
rs897323903 Chr.X: 53434340-53434340 on Build GRCh38
rs1003193515 Chr.X: 53434433-53434433 on Build GRCh38
rs782467682 Chr.X: 53434346-53434346 on Build GRCh38
rs112303075 Chr.X: 53434315-53434315 on Build GRCh38
rs782653444 Chr.X: 53434355-53434355 on Build GRCh38
rs1010778997 Chr.X: 53434460-53434460 on Build GRCh38
rs1054734279 Chr.X: 53434444-53434444 on Build GRCh38
rs781786018 Chr.X: 53434357-53434357 on Build GRCh38

View Full Product Details