Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70523781-70524036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMTN
Entrez Gene ID
401138
Gene Name
amelotin
Gene Aliases
UNQ689
UniGene
Hs.453069
Gene Chromosome Location
Chr.4: 70518572-70532743 on Build GRCh38
Transcripts
NM_001286731.1 NM_212557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3765146 c.204+1G>T p.? Chr.4: 70523934-70523934 on Build GRCh38
7203966 c.204+2T>C p.? Chr.4: 70523935-70523935 on Build GRCh38
7355388 c.139G>A p.V47I Chr.4: 70523868-70523868 on Build GRCh38
4125749 c.146C>A p.P49H Chr.4: 70523875-70523875 on Build GRCh38
734834 c.191C>A p.P64Q Chr.4: 70523920-70523920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770752683 Chr.4: 70523839-70523839 on Build GRCh38
rs762941998 Chr.4: 70523851-70523851 on Build GRCh38
rs557438220 Chr.4: 70523995-70523995 on Build GRCh38
rs772949388 Chr.4: 70523817-70523817 on Build GRCh38
rs932783642 Chr.4: 70523999-70523999 on Build GRCh38
rs761441656 Chr.4: 70523860-70523860 on Build GRCh38
rs760930175 Chr.4: 70523962-70523962 on Build GRCh38
rs746991039 Chr.4: 70523829-70523829 on Build GRCh38
rs752376041 Chr.4: 70523901-70523901 on Build GRCh38
rs891098622 Chr.4: 70523994-70523994 on Build GRCh38
rs1051305293 Chr.4: 70523987-70523987 on Build GRCh38
rs185440248 Chr.4: 70523892-70523892 on Build GRCh38
rs775680477 Chr.4: 70523844-70523844 on Build GRCh38
rs140471397 Chr.4: 70523870-70523870 on Build GRCh38
rs780982733 Chr.4: 70523936-70523936 on Build GRCh38
rs772389699 Chr.4: 70523953-70523953 on Build GRCh38
rs969834940 Chr.4: 70523922-70523922 on Build GRCh38
rs888689528 Chr.4: 70523832-70523832 on Build GRCh38
rs201144112 Chr.4: 70523866-70523866 on Build GRCh38
rs969283753 Chr.4: 70523833-70523833 on Build GRCh38
rs960377770 Chr.4: 70523849-70523849 on Build GRCh38
rs34803339 Chr.4: 70523877-70523877 on Build GRCh38
rs774730814 Chr.4: 70523862-70523862 on Build GRCh38
rs746750412 Chr.4: 70523915-70523915 on Build GRCh38
rs376704523 Chr.4: 70523948-70523948 on Build GRCh38
rs778327423 Chr.4: 70523883-70523883 on Build GRCh38
rs3912755 Chr.4: 70523952-70523952 on Build GRCh38
rs192563032 Chr.4: 70523868-70523868 on Build GRCh38
rs745764200 Chr.4: 70523938-70523938 on Build GRCh38
rs773689348 Chr.4: 70523958-70523958 on Build GRCh38
rs371411532 Chr.4: 70523940-70523940 on Build GRCh38
rs770967888 Chr.4: 70523928-70523928 on Build GRCh38
rs367591948 Chr.4: 70523865-70523865 on Build GRCh38
rs777594875 Chr.4: 70523912-70523912 on Build GRCh38
rs932886273 Chr.4: 70523985-70523985 on Build GRCh38
rs754305380 Chr.4: 70523975-70523975 on Build GRCh38
rs764472841 Chr.4: 70523857-70523857 on Build GRCh38
rs754496770 Chr.4: 70523880-70523880 on Build GRCh38
rs200838365 Chr.4: 70523974-70523974 on Build GRCh38
rs1023112589 Chr.4: 70523921-70523921 on Build GRCh38
rs750104626 Chr.4: 70523876-70523876 on Build GRCh38

View Full Product Details