Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99286785-99287058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1A
Entrez Gene ID
124
Gene Name
alcohol dehydrogenase 1A (class I), alpha polypeptide
Gene Aliases
ADH1
UniGene
Hs.654433
Gene Chromosome Location
Chr.4: 99276366-99291028 on Build GRCh38
Transcripts
NM_000667.3 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000667.3 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595164 c.213C>G p.A71A Chr.4: 99286896-99286896 on Build GRCh38
6595165 c.232G>T p.G78* Chr.4: 99286877-99286877 on Build GRCh38
5436705 c.256C>T p.P86S Chr.4: 99286853-99286853 on Build GRCh38
6882624 c.174G>A p.M58I Chr.4: 99286935-99286935 on Build GRCh38
2951274 c.134G>A p.G45E Chr.4: 99286975-99286975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779096083 Chr.4: 99286939-99286939 on Build GRCh38
rs750970851 Chr.4: 99287020-99287020 on Build GRCh38
rs372171102 Chr.4: 99286864-99286864 on Build GRCh38
rs191855460 Chr.4: 99286991-99286991 on Build GRCh38
rs1037760683 Chr.4: 99286930-99286930 on Build GRCh38
rs372298457 Chr.4: 99286940-99286940 on Build GRCh38
rs572305117 Chr.4: 99286990-99286990 on Build GRCh38
rs574630610 Chr.4: 99286830-99286830 on Build GRCh38
rs760558326 Chr.4: 99286868-99286868 on Build GRCh38
rs367795983 Chr.4: 99286896-99286896 on Build GRCh38
rs756522988 Chr.4: 99286898-99286898 on Build GRCh38
rs757130922 Chr.4: 99286890-99286890 on Build GRCh38
rs763459905 Chr.4: 99287001-99287001 on Build GRCh38
rs774347242 Chr.4: 99286850-99286850 on Build GRCh38
rs759278507 Chr.4: 99286987-99286987 on Build GRCh38
rs746307780 Chr.4: 99286823-99286823 on Build GRCh38
rs954950782 Chr.4: 99286997-99286997 on Build GRCh38
rs768754387 Chr.4: 99286848-99286848 on Build GRCh38
rs968461566 Chr.4: 99286955-99286955 on Build GRCh38
rs766069801 Chr.4: 99286872-99286872 on Build GRCh38
rs761751115 Chr.4: 99286858-99286858 on Build GRCh38
rs753554281 Chr.4: 99286873-99286873 on Build GRCh38
rs749528685 Chr.4: 99286926-99286926 on Build GRCh38
rs750869023 Chr.4: 99286895-99286895 on Build GRCh38
rs755133699 Chr.4: 99286937-99286937 on Build GRCh38
rs752311049 Chr.4: 99286884-99286884 on Build GRCh38
rs993439267 Chr.4: 99286974-99286974 on Build GRCh38
rs763784038 Chr.4: 99286911-99286912 on Build GRCh38
rs762598355 Chr.4: 99286996-99286996 on Build GRCh38
rs377108015 Chr.4: 99287023-99287023 on Build GRCh38
rs772057947 Chr.4: 99286863-99286863 on Build GRCh38
rs759074518 Chr.4: 99286877-99286877 on Build GRCh38
rs1001865521 Chr.4: 99286880-99286880 on Build GRCh38
rs560382418 Chr.4: 99286959-99286959 on Build GRCh38
rs770020081 Chr.4: 99286829-99286829 on Build GRCh38
rs367883723 Chr.4: 99286815-99286815 on Build GRCh38
rs773251361 Chr.4: 99286941-99286941 on Build GRCh38
rs1052370388 Chr.4: 99286824-99286824 on Build GRCh38
rs780240818 Chr.4: 99286905-99286905 on Build GRCh38
rs776225461 Chr.4: 99286960-99286960 on Build GRCh38
rs781760522 Chr.4: 99286819-99286819 on Build GRCh38
rs1006724795 Chr.4: 99286922-99286922 on Build GRCh38
rs942080707 Chr.4: 99286861-99286861 on Build GRCh38
rs934955691 Chr.4: 99286820-99286820 on Build GRCh38
rs749421127 Chr.4: 99286838-99286838 on Build GRCh38
rs746876988 Chr.4: 99286954-99286954 on Build GRCh38
rs141454408 Chr.4: 99286889-99286889 on Build GRCh38
rs61732105 Chr.4: 99286882-99286882 on Build GRCh38
rs75069921 Chr.4: 99286847-99286847 on Build GRCh38

View Full Product Details