Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109507939-109508168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
UniGene
Hs.726479
Gene Chromosome Location
Chr.1: 109504178-109509742 on Build GRCh38
Transcripts
NM_020703.2 XM_011541812.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3965692 c.950C>G p.T317R Chr.1: 109507963-109507963 on Build GRCh38
6507607 c.830C>T p.A277V Chr.1: 109508083-109508083 on Build GRCh38
4661384 c.858G>C p.L286F Chr.1: 109508055-109508055 on Build GRCh38
127306 c.918T>C p.N306N Chr.1: 109507995-109507995 on Build GRCh38
79491 c.795G>A p.L265L Chr.1: 109508118-109508118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755638606 Chr.1: 109507994-109507994 on Build GRCh38
rs745861797 Chr.1: 109507990-109507990 on Build GRCh38
rs564085384 Chr.1: 109507968-109507968 on Build GRCh38
rs781124413 Chr.1: 109507984-109507984 on Build GRCh38
rs140461975 Chr.1: 109508015-109508015 on Build GRCh38
rs779480021 Chr.1: 109508006-109508006 on Build GRCh38
rs748921792 Chr.1: 109508007-109508007 on Build GRCh38
rs1055715080 Chr.1: 109508070-109508070 on Build GRCh38
rs906853923 Chr.1: 109507992-109507992 on Build GRCh38
rs368080476 Chr.1: 109508120-109508120 on Build GRCh38
rs777513107 Chr.1: 109507976-109507976 on Build GRCh38
rs200644494 Chr.1: 109508106-109508106 on Build GRCh38
rs751314333 Chr.1: 109507978-109507978 on Build GRCh38
rs931784447 Chr.1: 109508064-109508064 on Build GRCh38
rs751791109 Chr.1: 109508138-109508138 on Build GRCh38
rs374249705 Chr.1: 109507961-109507961 on Build GRCh38
rs764249735 Chr.1: 109508099-109508099 on Build GRCh38
rs1001223291 Chr.1: 109508061-109508061 on Build GRCh38
rs758664850 Chr.1: 109507969-109507969 on Build GRCh38
rs777188107 Chr.1: 109508049-109508049 on Build GRCh38
rs775472564 Chr.1: 109508087-109508087 on Build GRCh38
rs763051198 Chr.1: 109508093-109508093 on Build GRCh38
rs757065041 Chr.1: 109507980-109507980 on Build GRCh38
rs779126263 Chr.1: 109508009-109508009 on Build GRCh38
rs760015516 Chr.1: 109508085-109508085 on Build GRCh38
rs765333109 Chr.1: 109508086-109508086 on Build GRCh38
rs35380866 Chr.1: 109508008-109508009 on Build GRCh38
rs771202155 Chr.1: 109508034-109508034 on Build GRCh38

View Full Product Details