Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11575063-11575292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP5
Entrez Gene ID
54
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
UniGene
Hs.1211
Gene Chromosome Location
Chr.19: 11574660-11579008 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Gene Symbol
ZNF627
Entrez Gene ID
199692
Gene Name
zinc finger protein 627
Gene Aliases
-
UniGene
Hs.526665
Gene Chromosome Location
Chr.19: 11575255-11619159 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5388624 c.924G>A p.E308E Chr.19: 11575064-11575064 on Build GRCh38
3730277 c.807G>A p.R269R Chr.19: 11575181-11575181 on Build GRCh38
6875492 c.824C>T p.P275L Chr.19: 11575164-11575164 on Build GRCh38
5758088 c.767T>G p.V256G Chr.19: 11575221-11575221 on Build GRCh38
4813585 c.742C>G p.Q248E Chr.19: 11575246-11575246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776546224 Chr.19: 11575233-11575233 on Build GRCh38
rs766031438 Chr.19: 11575259-11575259 on Build GRCh38
rs368442469 Chr.19: 11575211-11575211 on Build GRCh38
rs371116310 Chr.19: 11575231-11575231 on Build GRCh38
rs764873917 Chr.19: 11575252-11575252 on Build GRCh38
rs201276767 Chr.19: 11575119-11575119 on Build GRCh38
rs775399613 Chr.19: 11575267-11575268 on Build GRCh38
rs931749942 Chr.19: 11575144-11575144 on Build GRCh38
rs147115345 Chr.19: 11575127-11575127 on Build GRCh38
rs150107007 Chr.19: 11575173-11575173 on Build GRCh38
rs769748906 Chr.19: 11575188-11575188 on Build GRCh38
rs778409873 Chr.19: 11575100-11575100 on Build GRCh38
rs192632577 Chr.19: 11575215-11575215 on Build GRCh38
rs373526402 Chr.19: 11575122-11575122 on Build GRCh38
rs775386822 Chr.19: 11575230-11575230 on Build GRCh38
rs764042654 Chr.19: 11575170-11575170 on Build GRCh38
rs751295033 Chr.19: 11575270-11575270 on Build GRCh38
rs377528244 Chr.19: 11575159-11575159 on Build GRCh38
rs756856200 Chr.19: 11575088-11575088 on Build GRCh38
rs750130313 Chr.19: 11575107-11575107 on Build GRCh38
rs147025508 Chr.19: 11575174-11575174 on Build GRCh38
rs752091275 Chr.19: 11575117-11575117 on Build GRCh38
rs775646090 Chr.19: 11575161-11575161 on Build GRCh38
rs990482799 Chr.19: 11575087-11575087 on Build GRCh38
rs188463838 Chr.19: 11575186-11575186 on Build GRCh38
rs62638748 Chr.19: 11575128-11575128 on Build GRCh38
rs761798208 Chr.19: 11575250-11575250 on Build GRCh38
rs749645391 Chr.19: 11575142-11575142 on Build GRCh38
rs765765129 Chr.19: 11575193-11575193 on Build GRCh38
rs77911902 Chr.19: 11575133-11575133 on Build GRCh38
rs774428695 Chr.19: 11575150-11575150 on Build GRCh38
rs773090099 Chr.19: 11575255-11575255 on Build GRCh38
rs387906671 Chr.19: 11575155-11575157 on Build GRCh38
rs771458690 Chr.19: 11575149-11575149 on Build GRCh38
rs549955384 Chr.19: 11575223-11575223 on Build GRCh38
rs387906670 Chr.19: 11575197-11575197 on Build GRCh38
rs199986980 Chr.19: 11575183-11575183 on Build GRCh38
rs931778512 Chr.19: 11575140-11575140 on Build GRCh38
rs139096747 Chr.19: 11575232-11575232 on Build GRCh38
rs141519703 Chr.19: 11575205-11575205 on Build GRCh38
rs371003771 Chr.19: 11575160-11575160 on Build GRCh38
rs146196342 Chr.19: 11575222-11575222 on Build GRCh38
rs757680465 Chr.19: 11575218-11575218 on Build GRCh38
rs760569679 Chr.19: 11575163-11575163 on Build GRCh38
rs749985898 Chr.19: 11575182-11575182 on Build GRCh38
rs913182619 Chr.19: 11575116-11575116 on Build GRCh38
rs879255600 Chr.19: 11575171-11575172 on Build GRCh38
rs878853218 Chr.19: 11575198-11575216 on Build GRCh38

View Full Product Details