Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157491696-157491943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM19
Entrez Gene ID
8728
Gene Name
ADAM metallopeptidase domain 19
Gene Aliases
FKSG34, MADDAM, MLTNB
UniGene
Hs.483944
Gene Chromosome Location
Chr.5: 157477304-157575823 on Build GRCh38
Transcripts
NM_033274.4 XM_005266003.2 XM_011534682.2 XM_017010009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3716624 c.1993-4C>G p.? Chr.5: 157491727-157491727 on Build GRCh38
5770419 c.1954G>T p.E652* Chr.5: 157491873-157491873 on Build GRCh38
5770420 c.1147G>T p.E383* Chr.5: 157491873-157491873 on Build GRCh38
6227598 c.1108-7C>T p.? Chr.5: 157491919-157491919 on Build GRCh38
6170442 c.1932G>A p.R644R Chr.5: 157491889-157491889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755039969 Chr.5: 157491727-157491727 on Build GRCh38
rs200883455 Chr.5: 157491832-157491832 on Build GRCh38
rs771495678 Chr.5: 157491771-157491771 on Build GRCh38
rs372336872 Chr.5: 157491752-157491752 on Build GRCh38
rs201100159 Chr.5: 157491768-157491768 on Build GRCh38
rs763838612 Chr.5: 157491807-157491807 on Build GRCh38
rs775010615 Chr.5: 157491774-157491774 on Build GRCh38
rs2287749 Chr.5: 157491842-157491842 on Build GRCh38
rs746346343 Chr.5: 157491916-157491916 on Build GRCh38
rs749559925 Chr.5: 157491869-157491869 on Build GRCh38
rs762790571 Chr.5: 157491879-157491881 on Build GRCh38
rs921804703 Chr.5: 157491756-157491756 on Build GRCh38
rs749691225 Chr.5: 157491762-157491762 on Build GRCh38
rs746434083 Chr.5: 157491785-157491785 on Build GRCh38
rs561948234 Chr.5: 157491838-157491838 on Build GRCh38
rs752847811 Chr.5: 157491854-157491854 on Build GRCh38
rs964047306 Chr.5: 157491725-157491725 on Build GRCh38
rs374726786 Chr.5: 157491796-157491796 on Build GRCh38
rs777947434 Chr.5: 157491757-157491757 on Build GRCh38
rs182092067 Chr.5: 157491831-157491831 on Build GRCh38
rs778063164 Chr.5: 157491863-157491863 on Build GRCh38
rs146858323 Chr.5: 157491877-157491877 on Build GRCh38
rs140037727 Chr.5: 157491841-157491841 on Build GRCh38
rs760195529 Chr.5: 157491797-157491797 on Build GRCh38
rs752574745 Chr.5: 157491857-157491858 on Build GRCh38
rs985018271 Chr.5: 157491776-157491776 on Build GRCh38
rs748291556 Chr.5: 157491739-157491739 on Build GRCh38
rs952009424 Chr.5: 157491765-157491765 on Build GRCh38
rs758368854 Chr.5: 157491839-157491839 on Build GRCh38
rs771703929 Chr.5: 157491789-157491789 on Build GRCh38
rs369351955 Chr.5: 157491919-157491919 on Build GRCh38
rs772728570 Chr.5: 157491918-157491918 on Build GRCh38
rs756253549 Chr.5: 157491861-157491861 on Build GRCh38
rs141502864 Chr.5: 157491779-157491779 on Build GRCh38
rs368768876 Chr.5: 157491735-157491735 on Build GRCh38
rs947690198 Chr.5: 157491920-157491920 on Build GRCh38
rs765072237 Chr.5: 157491834-157491834 on Build GRCh38
rs779487366 Chr.5: 157491898-157491898 on Build GRCh38
rs11466791 Chr.5: 157491780-157491780 on Build GRCh38

View Full Product Details