Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20973930-20974173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4582226 c.357C>T p.H119H Chr.22: 20974100-20974100 on Build GRCh38
4582225 c.411C>T p.H137H Chr.22: 20974100-20974100 on Build GRCh38
6373375 c.379C>T p.R127C Chr.22: 20974086-20974086 on Build GRCh38
3552354 c.370G>A p.G124S Chr.22: 20974077-20974077 on Build GRCh38
5589594 c.385C>T p.P129S Chr.22: 20974092-20974092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768398726 Chr.22: 20974104-20974104 on Build GRCh38
rs749009581 Chr.22: 20974103-20974103 on Build GRCh38
rs981733554 Chr.22: 20974026-20974026 on Build GRCh38
rs769592838 Chr.22: 20974041-20974041 on Build GRCh38
rs374048838 Chr.22: 20974087-20974087 on Build GRCh38
rs753990478 Chr.22: 20974080-20974080 on Build GRCh38
rs915483669 Chr.22: 20974000-20974000 on Build GRCh38
rs763410536 Chr.22: 20974093-20974093 on Build GRCh38
rs371603801 Chr.22: 20974058-20974058 on Build GRCh38
rs373337824 Chr.22: 20974037-20974037 on Build GRCh38
rs768350092 Chr.22: 20974065-20974065 on Build GRCh38
rs760222060 Chr.22: 20973959-20973959 on Build GRCh38
rs375070516 Chr.22: 20974020-20974020 on Build GRCh38
rs755187029 Chr.22: 20974082-20974082 on Build GRCh38
rs981565799 Chr.22: 20973983-20973984 on Build GRCh38
rs542222592 Chr.22: 20974064-20974064 on Build GRCh38
rs892801584 Chr.22: 20973962-20973962 on Build GRCh38
rs201450835 Chr.22: 20974146-20974146 on Build GRCh38
rs971091166 Chr.22: 20973968-20973968 on Build GRCh38
rs376565725 Chr.22: 20973996-20973996 on Build GRCh38
rs918222551 Chr.22: 20973963-20973963 on Build GRCh38
rs538782836 Chr.22: 20974075-20974075 on Build GRCh38
rs368820001 Chr.22: 20974100-20974100 on Build GRCh38
rs757250012 Chr.22: 20974092-20974092 on Build GRCh38
rs750564830 Chr.22: 20974086-20974086 on Build GRCh38
rs779994284 Chr.22: 20974101-20974101 on Build GRCh38
rs370362252 Chr.22: 20974074-20974074 on Build GRCh38
rs773723658 Chr.22: 20974067-20974067 on Build GRCh38
rs1051028537 Chr.22: 20974132-20974132 on Build GRCh38
rs766601186 Chr.22: 20974079-20974079 on Build GRCh38
rs775229338 Chr.22: 20974047-20974047 on Build GRCh38
rs567610697 Chr.22: 20973997-20973997 on Build GRCh38
rs747033888 Chr.22: 20974025-20974025 on Build GRCh38
rs770515250 Chr.22: 20974140-20974141 on Build GRCh38
rs746636724 Chr.22: 20974023-20974023 on Build GRCh38
rs1011228695 Chr.22: 20973987-20973987 on Build GRCh38
rs369532939 Chr.22: 20974031-20974031 on Build GRCh38
rs753460407 Chr.22: 20974061-20974061 on Build GRCh38
rs927100374 Chr.22: 20973988-20973988 on Build GRCh38
rs760897659 Chr.22: 20974077-20974077 on Build GRCh38
rs80133973 Chr.22: 20974038-20974038 on Build GRCh38
rs981170318 Chr.22: 20974096-20974096 on Build GRCh38
rs555769378 Chr.22: 20974108-20974108 on Build GRCh38

View Full Product Details