Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44267433-44267694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604547 c.814C>T p.R272W Chr.11: 44267586-44267586 on Build GRCh38
6598961 c.800C>T p.A267V Chr.11: 44267600-44267600 on Build GRCh38
1475432 c.794G>A p.R265Q Chr.11: 44267606-44267606 on Build GRCh38
1475431 c.886C>T p.R296* Chr.11: 44267514-44267514 on Build GRCh38
5501743 c.852C>A p.H284Q Chr.11: 44267548-44267548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768535130 Chr.11: 44267581-44267581 on Build GRCh38
rs368050443 Chr.11: 44267579-44267579 on Build GRCh38
rs765208460 Chr.11: 44267606-44267606 on Build GRCh38
rs778373738 Chr.11: 44267518-44267518 on Build GRCh38
rs768402959 Chr.11: 44267655-44267655 on Build GRCh38
rs756296886 Chr.11: 44267627-44267627 on Build GRCh38
rs200169828 Chr.11: 44267631-44267631 on Build GRCh38
rs760942042 Chr.11: 44267670-44267670 on Build GRCh38
rs771851241 Chr.11: 44267565-44267565 on Build GRCh38
rs767623025 Chr.11: 44267666-44267666 on Build GRCh38
rs144961504 Chr.11: 44267513-44267513 on Build GRCh38
rs114123605 Chr.11: 44267462-44267462 on Build GRCh38
rs144440589 Chr.11: 44267555-44267555 on Build GRCh38
rs267606653 Chr.11: 44267607-44267607 on Build GRCh38
rs534239033 Chr.11: 44267672-44267672 on Build GRCh38
rs746702019 Chr.11: 44267650-44267650 on Build GRCh38
rs767748164 Chr.11: 44267620-44267620 on Build GRCh38
rs759643811 Chr.11: 44267665-44267665 on Build GRCh38
rs776349238 Chr.11: 44267657-44267657 on Build GRCh38
rs375808395 Chr.11: 44267608-44267608 on Build GRCh38
rs775524684 Chr.11: 44267667-44267667 on Build GRCh38
rs757938036 Chr.11: 44267638-44267638 on Build GRCh38
rs112019901 Chr.11: 44267471-44267471 on Build GRCh38
rs745388939 Chr.11: 44267634-44267634 on Build GRCh38
rs747971557 Chr.11: 44267659-44267659 on Build GRCh38
rs756421390 Chr.11: 44267516-44267516 on Build GRCh38
rs764143920 Chr.11: 44267472-44267472 on Build GRCh38
rs530368100 Chr.11: 44267569-44267569 on Build GRCh38
rs371742894 Chr.11: 44267584-44267584 on Build GRCh38
rs373589363 Chr.11: 44267500-44267500 on Build GRCh38
rs193290 Chr.11: 44267636-44267636 on Build GRCh38
rs377477011 Chr.11: 44267484-44267484 on Build GRCh38
rs104894196 Chr.11: 44267585-44267585 on Build GRCh38
rs766253750 Chr.11: 44267632-44267632 on Build GRCh38
rs75147697 Chr.11: 44267633-44267633 on Build GRCh38
rs764414424 Chr.11: 44267630-44267630 on Build GRCh38
rs759545911 Chr.11: 44267609-44267609 on Build GRCh38
rs376834139 Chr.11: 44267461-44267461 on Build GRCh38
rs752876582 Chr.11: 44267623-44267623 on Build GRCh38
rs770353383 Chr.11: 44267457-44267457 on Build GRCh38
rs746826513 Chr.11: 44267580-44267580 on Build GRCh38
rs745376185 Chr.11: 44267557-44267557 on Build GRCh38
rs748499687 Chr.11: 44267452-44267452 on Build GRCh38
rs202170828 Chr.11: 44267466-44267466 on Build GRCh38
rs753073089 Chr.11: 44267514-44267514 on Build GRCh38
rs770305162 Chr.11: 44267464-44267464 on Build GRCh38
rs771444101 Chr.11: 44267552-44267552 on Build GRCh38
rs761707439 Chr.11: 44267602-44267602 on Build GRCh38
rs766411205 Chr.11: 44267499-44267499 on Build GRCh38
rs779886316 Chr.11: 44267643-44267643 on Build GRCh38
rs764486762 Chr.11: 44267570-44267570 on Build GRCh38
rs769624652 Chr.11: 44267663-44267663 on Build GRCh38
rs370265770 Chr.11: 44267485-44267485 on Build GRCh38
rs12419361 Chr.11: 44267521-44267521 on Build GRCh38
rs773879165 Chr.11: 44267465-44267465 on Build GRCh38

View Full Product Details