Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3753964-3754211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA3
Entrez Gene ID
9546
Gene Name
amyloid beta precursor protein binding family A member 3
Gene Aliases
MGC:15815, X11L2, mint3
UniGene
Hs.25527
Gene Chromosome Location
Chr.19: 3750773-3761697 on Build GRCh38
Transcripts
NM_004886.3 XM_006722950.3 XM_006722951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362822 c.846C>G p.S282S Chr.19: 3754022-3754022 on Build GRCh38
5517224 c.756C>T p.R252R Chr.19: 3754201-3754201 on Build GRCh38
6351229 c.754C>T p.R252C Chr.19: 3754203-3754203 on Build GRCh38
5601028 c.766C>T p.P256S Chr.19: 3754102-3754102 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs565229318 Chr.19: 3754061-3754061 on Build GRCh38
rs983581481 Chr.19: 3754020-3754020 on Build GRCh38
rs575937132 Chr.19: 3754174-3754174 on Build GRCh38
rs751346469 Chr.19: 3754120-3754120 on Build GRCh38
rs756881735 Chr.19: 3754014-3754014 on Build GRCh38
rs767228548 Chr.19: 3754006-3754006 on Build GRCh38
rs367758435 Chr.19: 3754001-3754001 on Build GRCh38
rs777853566 Chr.19: 3754156-3754156 on Build GRCh38
rs748717776 Chr.19: 3753998-3753998 on Build GRCh38
rs750309502 Chr.19: 3754029-3754029 on Build GRCh38
rs776507332 Chr.19: 3753996-3753996 on Build GRCh38
rs200609002 Chr.19: 3754054-3754054 on Build GRCh38
rs372589212 Chr.19: 3754117-3754117 on Build GRCh38
rs751835319 Chr.19: 3754040-3754040 on Build GRCh38
rs780418293 Chr.19: 3753984-3753984 on Build GRCh38
rs1025136378 Chr.19: 3754166-3754167 on Build GRCh38
rs774635160 Chr.19: 3754077-3754077 on Build GRCh38
rs139904607 Chr.19: 3754073-3754073 on Build GRCh38
rs770149370 Chr.19: 3753999-3753999 on Build GRCh38
rs747143405 Chr.19: 3753988-3753988 on Build GRCh38
rs143355076 Chr.19: 3754076-3754076 on Build GRCh38
rs752886270 Chr.19: 3754127-3754127 on Build GRCh38
rs746210138 Chr.19: 3754166-3754166 on Build GRCh38
rs3746119 Chr.19: 3754041-3754041 on Build GRCh38
rs370070756 Chr.19: 3754018-3754018 on Build GRCh38
rs904330782 Chr.19: 3754132-3754132 on Build GRCh38
rs750397386 Chr.19: 3754116-3754116 on Build GRCh38
rs375481797 Chr.19: 3754119-3754119 on Build GRCh38
rs539290720 Chr.19: 3754170-3754170 on Build GRCh38
rs368113227 Chr.19: 3754126-3754126 on Build GRCh38
rs760353050 Chr.19: 3754079-3754079 on Build GRCh38
rs767841379 Chr.19: 3754188-3754188 on Build GRCh38
rs781350702 Chr.19: 3754051-3754051 on Build GRCh38
rs748352093 Chr.19: 3754031-3754031 on Build GRCh38
rs749804214 Chr.19: 3754159-3754159 on Build GRCh38
rs772808248 Chr.19: 3754175-3754175 on Build GRCh38
rs146035497 Chr.19: 3754060-3754060 on Build GRCh38
rs142650185 Chr.19: 3754039-3754039 on Build GRCh38
rs770370823 Chr.19: 3754055-3754055 on Build GRCh38
rs763269469 Chr.19: 3754002-3754002 on Build GRCh38
rs897937074 Chr.19: 3754143-3754143 on Build GRCh38
rs527680633 Chr.19: 3754163-3754163 on Build GRCh38
rs768868526 Chr.19: 3753989-3753989 on Build GRCh38
rs548325977 Chr.19: 3754171-3754171 on Build GRCh38
rs775115362 Chr.19: 3754015-3754015 on Build GRCh38
rs890201932 Chr.19: 3754099-3754099 on Build GRCh38
rs765275684 Chr.19: 3754114-3754114 on Build GRCh38
rs541683528 Chr.19: 3754184-3754184 on Build GRCh38
rs370561473 Chr.19: 3754113-3754113 on Build GRCh38
rs61731066 Chr.19: 3754022-3754022 on Build GRCh38
rs781581796 Chr.19: 3754173-3754182 on Build GRCh38
rs752877886 Chr.19: 3754191-3754191 on Build GRCh38
rs1000134597 Chr.19: 3754153-3754153 on Build GRCh38
rs368597663 Chr.19: 3754104-3754104 on Build GRCh38
rs760267251 Chr.19: 3754016-3754016 on Build GRCh38
rs763003232 Chr.19: 3754180-3754180 on Build GRCh38
rs765064793 Chr.19: 3754028-3754028 on Build GRCh38
rs371745996 Chr.19: 3754162-3754162 on Build GRCh38
rs561957330 Chr.19: 3754185-3754185 on Build GRCh38
rs756137161 Chr.19: 3754129-3754129 on Build GRCh38
rs374472856 Chr.19: 3753982-3753982 on Build GRCh38
rs768144635 Chr.19: 3754100-3754100 on Build GRCh38

View Full Product Details