Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219232253-219232512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKZF1
Entrez Gene ID
55139
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
ZNF744
UniGene
Hs.437647
Gene Chromosome Location
Chr.2: 219229757-219236669 on Build GRCh38
Transcripts
NM_001042410.1 NM_001282792.1 NM_018089.2 XM_005246663.2 XM_011511392.2 XM_017004420.1 XM_017004421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4091639 c.383C>T p.S128L Chr.2: 219232508-219232508 on Build GRCh38
4661990 c.330G>A p.L110L Chr.2: 219232328-219232328 on Build GRCh38
1016646 c.359C>T p.S120F Chr.2: 219232357-219232357 on Build GRCh38
6600972 c.351G>A p.K117K Chr.2: 219232349-219232349 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764832677 Chr.2: 219232287-219232287 on Build GRCh38
rs753627625 Chr.2: 219232388-219232388 on Build GRCh38
rs761017394 Chr.2: 219232288-219232288 on Build GRCh38
rs776690007 Chr.2: 219232304-219232304 on Build GRCh38
rs200147041 Chr.2: 219232330-219232330 on Build GRCh38
rs375545713 Chr.2: 219232463-219232463 on Build GRCh38
rs905054947 Chr.2: 219232397-219232397 on Build GRCh38
rs771433543 Chr.2: 219232476-219232476 on Build GRCh38
rs747415610 Chr.2: 219232343-219232343 on Build GRCh38
rs772627468 Chr.2: 219232340-219232340 on Build GRCh38
rs576741102 Chr.2: 219232305-219232305 on Build GRCh38
rs750350626 Chr.2: 219232385-219232385 on Build GRCh38
rs778567947 Chr.2: 219232443-219232443 on Build GRCh38
rs767507073 Chr.2: 219232313-219232313 on Build GRCh38
rs746189242 Chr.2: 219232484-219232484 on Build GRCh38
rs765254854 Chr.2: 219232306-219232306 on Build GRCh38
rs755965392 Chr.2: 219232316-219232316 on Build GRCh38
rs878974266 Chr.2: 219232447-219232447 on Build GRCh38
rs764262384 Chr.2: 219232488-219232490 on Build GRCh38
rs572744276 Chr.2: 219232386-219232386 on Build GRCh38
rs777010161 Chr.2: 219232359-219232359 on Build GRCh38
rs747589774 Chr.2: 219232336-219232336 on Build GRCh38
rs778316061 Chr.2: 219232332-219232332 on Build GRCh38
rs191253543 Chr.2: 219232378-219232378 on Build GRCh38
rs766383130 Chr.2: 219232488-219232488 on Build GRCh38
rs367763893 Chr.2: 219232377-219232377 on Build GRCh38
rs767625985 Chr.2: 219232380-219232380 on Build GRCh38
rs771177890 Chr.2: 219232349-219232349 on Build GRCh38
rs748865889 Chr.2: 219232325-219232325 on Build GRCh38
rs779792670 Chr.2: 219232323-219232323 on Build GRCh38
rs757874315 Chr.2: 219232448-219232448 on Build GRCh38
rs539189014 Chr.2: 219232289-219232289 on Build GRCh38
rs879192784 Chr.2: 219232350-219232350 on Build GRCh38
rs752784056 Chr.2: 219232310-219232310 on Build GRCh38
rs370626225 Chr.2: 219232477-219232477 on Build GRCh38
rs754609259 Chr.2: 219232328-219232328 on Build GRCh38
rs754589734 Chr.2: 219232400-219232400 on Build GRCh38
rs750299159 Chr.2: 219232314-219232314 on Build GRCh38
rs541348687 Chr.2: 219232431-219232431 on Build GRCh38
rs956418928 Chr.2: 219232489-219232489 on Build GRCh38
rs778301329 Chr.2: 219232460-219232460 on Build GRCh38
rs761254387 Chr.2: 219232363-219232363 on Build GRCh38
rs199508320 Chr.2: 219232490-219232490 on Build GRCh38

View Full Product Details