Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201707798-201708071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2
Entrez Gene ID
57679
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Gene Aliases
ALS2CR6, ALSJ, IAHSP, PLSJ
UniGene
Hs.471096
Gene Chromosome Location
Chr.2: 201700263-201781172 on Build GRCh38
Transcripts
NM_020919.3 XM_006712654.2 XM_006712655.3 XM_011511530.2 XM_017004569.1 XM_017004571.1 XM_017004572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
359618 c.4303G>A p.E1435K Chr.2: 201707969-201707969 on Build GRCh38
1014953 c.4382G>A p.R1461Q Chr.2: 201707890-201707890 on Build GRCh38
5015971 c.4368delG p.K1457fs*14 Chr.2: 201707904-201707904 on Build GRCh38
6466836 c.4398G>A p.E1466E Chr.2: 201707874-201707874 on Build GRCh38
3724630 c.4328C>G p.S1443C Chr.2: 201707944-201707944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758889641 Chr.2: 201707890-201707890 on Build GRCh38
rs771938136 Chr.2: 201707858-201707858 on Build GRCh38
rs927679453 Chr.2: 201707909-201707909 on Build GRCh38
rs561437381 Chr.2: 201707985-201707985 on Build GRCh38
rs568284468 Chr.2: 201707973-201707973 on Build GRCh38
rs779859483 Chr.2: 201707836-201707836 on Build GRCh38
rs757764512 Chr.2: 201707905-201707905 on Build GRCh38
rs762948016 Chr.2: 201708026-201708026 on Build GRCh38
rs939110770 Chr.2: 201707935-201707935 on Build GRCh38
rs886055452 Chr.2: 201707906-201707906 on Build GRCh38
rs747637647 Chr.2: 201707967-201707967 on Build GRCh38
rs755770548 Chr.2: 201707834-201707834 on Build GRCh38
rs376011573 Chr.2: 201707955-201707955 on Build GRCh38
rs1031101585 Chr.2: 201708037-201708037 on Build GRCh38
rs766580828 Chr.2: 201707864-201707864 on Build GRCh38
rs760463574 Chr.2: 201707863-201707863 on Build GRCh38
rs180758472 Chr.2: 201707873-201707873 on Build GRCh38
rs772998097 Chr.2: 201707860-201707860 on Build GRCh38
rs781648792 Chr.2: 201707912-201707914 on Build GRCh38
rs755283897 Chr.2: 201707854-201707854 on Build GRCh38
rs748999549 Chr.2: 201707928-201707928 on Build GRCh38
rs765396371 Chr.2: 201707886-201707886 on Build GRCh38
rs374047961 Chr.2: 201707891-201707891 on Build GRCh38
rs775582606 Chr.2: 201708025-201708025 on Build GRCh38
rs746790680 Chr.2: 201707991-201707991 on Build GRCh38
rs779841299 Chr.2: 201707927-201707927 on Build GRCh38
rs1005432754 Chr.2: 201707929-201707929 on Build GRCh38
rs765399760 Chr.2: 201708023-201708023 on Build GRCh38
rs759713373 Chr.2: 201708020-201708020 on Build GRCh38
rs747165744 Chr.2: 201707853-201707853 on Build GRCh38
rs748803335 Chr.2: 201707842-201707842 on Build GRCh38
rs894178402 Chr.2: 201707946-201707946 on Build GRCh38
rs752751086 Chr.2: 201707888-201707888 on Build GRCh38
rs571161154 Chr.2: 201707843-201707843 on Build GRCh38
rs538132017 Chr.2: 201707957-201707957 on Build GRCh38
rs888377361 Chr.2: 201707902-201707902 on Build GRCh38
rs768037268 Chr.2: 201708036-201708036 on Build GRCh38
rs748161670 Chr.2: 201707853-201707853 on Build GRCh38
rs752172105 Chr.2: 201708032-201708032 on Build GRCh38
rs764523082 Chr.2: 201707897-201707897 on Build GRCh38
rs762249415 Chr.2: 201708035-201708035 on Build GRCh38
rs759588722 Chr.2: 201707881-201707881 on Build GRCh38
rs751906518 Chr.2: 201707904-201707904 on Build GRCh38
rs764659467 Chr.2: 201708027-201708027 on Build GRCh38
rs1000224325 Chr.2: 201708033-201708033 on Build GRCh38
rs773196767 Chr.2: 201707981-201707981 on Build GRCh38
rs549986860 Chr.2: 201707968-201707968 on Build GRCh38
rs775146442 Chr.2: 201707846-201707846 on Build GRCh38
rs776801576 Chr.2: 201708015-201708015 on Build GRCh38
rs370365603 Chr.2: 201707876-201707876 on Build GRCh38
rs371872969 Chr.2: 201708011-201708011 on Build GRCh38

View Full Product Details