Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56353617-56353888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFN1
Entrez Gene ID
162282
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
-
UniGene
Hs.673040
Gene Chromosome Location
Chr.17: 56110935-56517016 on Build GRCh38
Transcripts
NM_153228.2 XM_006721728.3 XM_011524428.2 XM_011524429.2 XM_011524430.2 XM_011524431.2 XM_017024263.1 XM_017024264.1 XM_017024265.1 XM_017024266.1 XM_017024267.1 XM_017024268.1 XM_017024269.1 XM_017024270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750177 c.427G>C p.E143Q Chr.17: 56353863-56353863 on Build GRCh38
5933522 c.400-8C>T p.? Chr.17: 56353828-56353828 on Build GRCh38
5043271 c.450G>T p.M150I Chr.17: 56353886-56353886 on Build GRCh38
981605 c.400A>C p.N134H Chr.17: 56353836-56353836 on Build GRCh38
4571208 c.404T>A p.F135Y Chr.17: 56353840-56353840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564431912 Chr.17: 56353699-56353699 on Build GRCh38
rs926535076 Chr.17: 56353779-56353779 on Build GRCh38
rs970422004 Chr.17: 56353705-56353705 on Build GRCh38
rs1011463812 Chr.17: 56353709-56353709 on Build GRCh38
rs1020652124 Chr.17: 56353711-56353711 on Build GRCh38
rs775647626 Chr.17: 56353824-56353824 on Build GRCh38
rs1035493116 Chr.17: 56353727-56353727 on Build GRCh38
rs749853257 Chr.17: 56353828-56353828 on Build GRCh38
rs747187408 Chr.17: 56353680-56353681 on Build GRCh38
rs958011212 Chr.17: 56353648-56353648 on Build GRCh38
rs892782099 Chr.17: 56353664-56353664 on Build GRCh38
rs981986567 Chr.17: 56353765-56353765 on Build GRCh38
rs903084968 Chr.17: 56353722-56353722 on Build GRCh38
rs768185507 Chr.17: 56353863-56353863 on Build GRCh38
rs748234868 Chr.17: 56353813-56353813 on Build GRCh38
rs774605535 Chr.17: 56353843-56353843 on Build GRCh38
rs144270571 Chr.17: 56353796-56353796 on Build GRCh38
rs1026999503 Chr.17: 56353815-56353815 on Build GRCh38
rs747203592 Chr.17: 56353793-56353793 on Build GRCh38
rs1003151354 Chr.17: 56353723-56353723 on Build GRCh38
rs779743953 Chr.17: 56353792-56353792 on Build GRCh38
rs546102964 Chr.17: 56353682-56353682 on Build GRCh38
rs148744090 Chr.17: 56353859-56353859 on Build GRCh38
rs550966854 Chr.17: 56353825-56353825 on Build GRCh38
rs771290487 Chr.17: 56353833-56353833 on Build GRCh38
rs770056732 Chr.17: 56353821-56353821 on Build GRCh38
rs775163894 Chr.17: 56353663-56353663 on Build GRCh38
rs369696032 Chr.17: 56353706-56353706 on Build GRCh38
rs781265797 Chr.17: 56353800-56353800 on Build GRCh38

View Full Product Details