Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69250449-69250688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA5
Entrez Gene ID
23461
Gene Name
ATP binding cassette subfamily A member 5
Gene Aliases
ABC13, EST90625
UniGene
Hs.731824
Gene Chromosome Location
Chr.17: 69244435-69327182 on Build GRCh38
Transcripts
NM_018672.4 NM_172232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589778 c.4643G>A p.R1548K Chr.17: 69250514-69250514 on Build GRCh38
1385569 c.4589delT p.L1530fs*5 Chr.17: 69250568-69250568 on Build GRCh38
6465118 c.4668T>A p.N1556K Chr.17: 69250489-69250489 on Build GRCh38
5564565 c.4676G>T p.R1559L Chr.17: 69250481-69250481 on Build GRCh38
5672663 c.4536-2A>G p.? Chr.17: 69250623-69250623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761451205 Chr.17: 69250604-69250604 on Build GRCh38
rs369801152 Chr.17: 69250494-69250494 on Build GRCh38
rs376917230 Chr.17: 69250589-69250589 on Build GRCh38
rs1007574033 Chr.17: 69250548-69250548 on Build GRCh38
rs781240794 Chr.17: 69250529-69250529 on Build GRCh38
rs369153660 Chr.17: 69250591-69250591 on Build GRCh38
rs559974558 Chr.17: 69250482-69250482 on Build GRCh38
rs527916132 Chr.17: 69250525-69250525 on Build GRCh38
rs375287029 Chr.17: 69250653-69250653 on Build GRCh38
rs764655925 Chr.17: 69250606-69250606 on Build GRCh38
rs571588399 Chr.17: 69250624-69250624 on Build GRCh38
rs752011696 Chr.17: 69250615-69250615 on Build GRCh38
rs899788359 Chr.17: 69250559-69250559 on Build GRCh38
rs761202660 Chr.17: 69250498-69250498 on Build GRCh38
rs750887568 Chr.17: 69250508-69250508 on Build GRCh38
rs758907008 Chr.17: 69250514-69250514 on Build GRCh38
rs766953080 Chr.17: 69250614-69250614 on Build GRCh38
rs763575719 Chr.17: 69250608-69250608 on Build GRCh38
rs761160032 Chr.17: 69250582-69250582 on Build GRCh38
rs765828096 Chr.17: 69250506-69250506 on Build GRCh38
rs148561229 Chr.17: 69250481-69250481 on Build GRCh38
rs746099868 Chr.17: 69250641-69250641 on Build GRCh38
rs768399602 Chr.17: 69250594-69250594 on Build GRCh38
rs749320449 Chr.17: 69250658-69250658 on Build GRCh38
rs375026872 Chr.17: 69250586-69250586 on Build GRCh38
rs79140751 Chr.17: 69250636-69250637 on Build GRCh38
rs141978506 Chr.17: 69250573-69250573 on Build GRCh38
rs567661810 Chr.17: 69250572-69250572 on Build GRCh38
rs779369766 Chr.17: 69250554-69250555 on Build GRCh38
rs118115378 Chr.17: 69250533-69250533 on Build GRCh38
rs758367117 Chr.17: 69250659-69250659 on Build GRCh38
rs778978598 Chr.17: 69250545-69250545 on Build GRCh38
rs369841294 Chr.17: 69250657-69250657 on Build GRCh38
rs201600235 Chr.17: 69250602-69250602 on Build GRCh38
rs370515109 Chr.17: 69250523-69250523 on Build GRCh38
rs186243305 Chr.17: 69250600-69250600 on Build GRCh38
rs781536653 Chr.17: 69250636-69250636 on Build GRCh38
rs1018197698 Chr.17: 69250547-69250547 on Build GRCh38
rs752859587 Chr.17: 69250637-69250637 on Build GRCh38
rs200779886 Chr.17: 69250524-69250524 on Build GRCh38

View Full Product Details