Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 32545810-32546067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP6
Entrez Gene ID
9472
Gene Name
A-kinase anchoring protein 6
Gene Aliases
ADAP100, ADAP6, AKAP100, PRKA6, mAKAP
UniGene
Hs.509083
Gene Chromosome Location
Chr.14: 32329273-32833999 on Build GRCh38
Transcripts
NM_004274.4 XM_017021808.1 XM_017021809.1 XM_017021810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7077959 c.1199C>G p.T400S Chr.14: 32545852-32545852 on Build GRCh38
955363 c.1254delG p.K418fs*4 Chr.14: 32545907-32545907 on Build GRCh38
3419773 c.1210G>T p.D404Y Chr.14: 32545863-32545863 on Build GRCh38
697958 c.1254G>T p.K418N Chr.14: 32545907-32545907 on Build GRCh38
79481 c.1323G>A p.L441L Chr.14: 32545976-32545976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200656320 Chr.14: 32545962-32545962 on Build GRCh38
rs777699344 Chr.14: 32545994-32545994 on Build GRCh38
rs112823706 Chr.14: 32545944-32545944 on Build GRCh38
rs372195655 Chr.14: 32545937-32545937 on Build GRCh38
rs752675928 Chr.14: 32545989-32545989 on Build GRCh38
rs776923297 Chr.14: 32546028-32546028 on Build GRCh38
rs776072608 Chr.14: 32545869-32545874 on Build GRCh38
rs923680463 Chr.14: 32545875-32545875 on Build GRCh38
rs777171525 Chr.14: 32545909-32545909 on Build GRCh38
rs142519432 Chr.14: 32546031-32546031 on Build GRCh38
rs973186374 Chr.14: 32545889-32545889 on Build GRCh38
rs769031091 Chr.14: 32546015-32546015 on Build GRCh38
rs780890969 Chr.14: 32545974-32545974 on Build GRCh38
rs762265218 Chr.14: 32545918-32545918 on Build GRCh38
rs771014636 Chr.14: 32545873-32545873 on Build GRCh38
rs781138670 Chr.14: 32545862-32545862 on Build GRCh38
rs754763453 Chr.14: 32545834-32545834 on Build GRCh38
rs756102728 Chr.14: 32545993-32545993 on Build GRCh38
rs750007249 Chr.14: 32545934-32545934 on Build GRCh38
rs1046662854 Chr.14: 32545868-32545868 on Build GRCh38
rs1039277621 Chr.14: 32545933-32545933 on Build GRCh38
rs764832400 Chr.14: 32545930-32545930 on Build GRCh38
rs761328092 Chr.14: 32546002-32546004 on Build GRCh38
rs571728105 Chr.14: 32546011-32546011 on Build GRCh38
rs911832733 Chr.14: 32545979-32545979 on Build GRCh38
rs1053427377 Chr.14: 32545902-32545902 on Build GRCh38
rs145516811 Chr.14: 32545941-32545941 on Build GRCh38
rs551902426 Chr.14: 32545888-32545888 on Build GRCh38
rs375052169 Chr.14: 32545896-32545896 on Build GRCh38
rs368322682 Chr.14: 32545907-32545907 on Build GRCh38
rs747241071 Chr.14: 32546013-32546013 on Build GRCh38
rs571042271 Chr.14: 32545900-32545900 on Build GRCh38
rs17099240 Chr.14: 32545876-32545876 on Build GRCh38
rs949245645 Chr.14: 32545833-32545833 on Build GRCh38
rs748087074 Chr.14: 32545863-32545863 on Build GRCh38
rs772168542 Chr.14: 32546007-32546007 on Build GRCh38

View Full Product Details