Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 15418565-15418834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGMO
Entrez Gene ID
392636
Gene Name
alkylglycerol monooxygenase
Gene Aliases
TMEM195
UniGene
Hs.670634
Gene Chromosome Location
Chr.7: 15117223-15562015 on Build GRCh38
Transcripts
NM_001004320.1 XM_006715730.1 XM_006715731.3 XM_011515402.2 XM_017012204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596289 c.518delT p.F173fs*41 Chr.7: 15418649-15418649 on Build GRCh38
3305123 c.519C>T p.F173F Chr.7: 15418648-15418648 on Build GRCh38
1673032 c.596G>T p.W199L Chr.7: 15418571-15418571 on Build GRCh38
7213970 c.542T>A p.I181K Chr.7: 15418625-15418625 on Build GRCh38
745773 c.547C>A p.P183T Chr.7: 15418620-15418620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753031361 Chr.7: 15418646-15418646 on Build GRCh38
rs760761781 Chr.7: 15418632-15418632 on Build GRCh38
rs973379858 Chr.7: 15418716-15418716 on Build GRCh38
rs545098769 Chr.7: 15418766-15418766 on Build GRCh38
rs769446617 Chr.7: 15418678-15418678 on Build GRCh38
rs754412614 Chr.7: 15418639-15418639 on Build GRCh38
rs766809607 Chr.7: 15418635-15418635 on Build GRCh38
rs916632370 Chr.7: 15418750-15418750 on Build GRCh38
rs1055216979 Chr.7: 15418743-15418743 on Build GRCh38
rs373210948 Chr.7: 15418606-15418606 on Build GRCh38
rs140890749 Chr.7: 15418737-15418737 on Build GRCh38
rs777374176 Chr.7: 15418599-15418599 on Build GRCh38
rs767404582 Chr.7: 15418619-15418619 on Build GRCh38
rs951977733 Chr.7: 15418668-15418668 on Build GRCh38
rs758015123 Chr.7: 15418648-15418648 on Build GRCh38
rs28754437 Chr.7: 15418705-15418705 on Build GRCh38
rs749060239 Chr.7: 15418700-15418700 on Build GRCh38
rs928018014 Chr.7: 15418734-15418734 on Build GRCh38
rs773104971 Chr.7: 15418620-15418620 on Build GRCh38
rs762137563 Chr.7: 15418623-15418623 on Build GRCh38
rs368209618 Chr.7: 15418775-15418775 on Build GRCh38
rs903220392 Chr.7: 15418758-15418758 on Build GRCh38
rs775316598 Chr.7: 15418682-15418682 on Build GRCh38
rs765849227 Chr.7: 15418668-15418669 on Build GRCh38
rs746452635 Chr.7: 15418658-15418658 on Build GRCh38
rs756892343 Chr.7: 15418661-15418661 on Build GRCh38
rs1040418032 Chr.7: 15418714-15418714 on Build GRCh38
rs192413362 Chr.7: 15418605-15418605 on Build GRCh38
rs777578023 Chr.7: 15418652-15418652 on Build GRCh38
rs760159093 Chr.7: 15418669-15418669 on Build GRCh38
rs199947500 Chr.7: 15418641-15418641 on Build GRCh38
rs746536370 Chr.7: 15418600-15418600 on Build GRCh38
rs765670837 Chr.7: 15418643-15418643 on Build GRCh38
rs373454548 Chr.7: 15418659-15418659 on Build GRCh38
rs573435174 Chr.7: 15418669-15418669 on Build GRCh38
rs750384516 Chr.7: 15418660-15418661 on Build GRCh38
rs767663830 Chr.7: 15418627-15418627 on Build GRCh38
rs923064944 Chr.7: 15418715-15418715 on Build GRCh38
rs1042886217 Chr.7: 15418774-15418774 on Build GRCh38
rs948545556 Chr.7: 15418771-15418771 on Build GRCh38
rs1002842264 Chr.7: 15418764-15418764 on Build GRCh38
rs1041932594 Chr.7: 15418752-15418752 on Build GRCh38
rs931682561 Chr.7: 15418727-15418727 on Build GRCh38
rs773227617 Chr.7: 15418628-15418628 on Build GRCh38
rs770933571 Chr.7: 15418603-15418603 on Build GRCh38

View Full Product Details