Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38367506-38367779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERG
Entrez Gene ID
2078
Gene Name
ERG, ETS transcription factor
Gene Aliases
erg-3, p55
UniGene
Hs.473819
Gene Chromosome Location
Chr.21: 38367261-38662045 on Build GRCh38
Transcripts
NM_001243432.2

SNP Detail

SNP ID Chromosome Location
rs2836346 Chr.21: 38367743-38367743 on Build GRCh38
rs896016141 Chr.21: 38367702-38367702 on Build GRCh38
rs146217369 Chr.21: 38367706-38367706 on Build GRCh38
rs939803114 Chr.21: 38367686-38367690 on Build GRCh38
rs557498207 Chr.21: 38367654-38367654 on Build GRCh38
rs959701579 Chr.21: 38367620-38367620 on Build GRCh38
rs760801356 Chr.21: 38367682-38367682 on Build GRCh38
rs991167393 Chr.21: 38367631-38367631 on Build GRCh38
rs1048397262 Chr.21: 38367665-38367665 on Build GRCh38
rs767504340 Chr.21: 38367555-38367555 on Build GRCh38
rs187145928 Chr.21: 38367678-38367678 on Build GRCh38
rs1041143598 Chr.21: 38367573-38367573 on Build GRCh38
rs533795413 Chr.21: 38367673-38367673 on Build GRCh38
rs953306992 Chr.21: 38367547-38367547 on Build GRCh38
rs901258048 Chr.21: 38367707-38367707 on Build GRCh38
rs918592675 Chr.21: 38367595-38367595 on Build GRCh38
rs925825293 Chr.21: 38367544-38367544 on Build GRCh38
rs376417021 Chr.21: 38367562-38367562 on Build GRCh38
rs114419699 Chr.21: 38367563-38367563 on Build GRCh38
rs868694191 Chr.21: 38367534-38367534 on Build GRCh38
rs759411459 Chr.21: 38367602-38367602 on Build GRCh38
rs1011519152 Chr.21: 38367554-38367554 on Build GRCh38
rs571208714 Chr.21: 38367672-38367672 on Build GRCh38
rs573061215 Chr.21: 38367726-38367726 on Build GRCh38
rs978700604 Chr.21: 38367531-38367531 on Build GRCh38
rs1029034078 Chr.21: 38367538-38367538 on Build GRCh38
rs997291378 Chr.21: 38367710-38367710 on Build GRCh38
rs767382517 Chr.21: 38367606-38367606 on Build GRCh38
rs1049735121 Chr.21: 38367714-38367714 on Build GRCh38
rs62217874 Chr.21: 38367556-38367556 on Build GRCh38
rs915843201 Chr.21: 38367632-38367632 on Build GRCh38
rs537681232 Chr.21: 38367601-38367601 on Build GRCh38

View Full Product Details