Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132827918-132828177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK8
Entrez Gene ID
158067
Gene Name
adenylate kinase 8
Gene Aliases
AK 8, C9orf98, DDX31
UniGene
Hs.421340
Gene Chromosome Location
Chr.9: 132725574-132878811 on Build GRCh38
Transcripts
NM_001317958.1 NM_001317959.1 NM_152572.2 XM_005272169.2 XM_006716965.2 XM_011518277.2 XM_011518278.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5552932 c.517G>A p.E173K Chr.9: 132828052-132828052 on Build GRCh38
4844912 c.547C>G p.Q183E Chr.9: 132828022-132828022 on Build GRCh38
1106310 c.521G>T p.R174I Chr.9: 132828048-132828048 on Build GRCh38
3788197 c.556+10C>T p.? Chr.9: 132828003-132828003 on Build GRCh38
7354988 c.537A>C p.R179S Chr.9: 132828032-132828032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763273973 Chr.9: 132827962-132827962 on Build GRCh38
rs774030270 Chr.9: 132827980-132827980 on Build GRCh38
rs1012518968 Chr.9: 132828029-132828029 on Build GRCh38
rs753293837 Chr.9: 132828129-132828129 on Build GRCh38
rs760438331 Chr.9: 132827986-132827986 on Build GRCh38
rs907917176 Chr.9: 132828141-132828141 on Build GRCh38
rs576117467 Chr.9: 132827950-132827950 on Build GRCh38
rs1057040252 Chr.9: 132828025-132828025 on Build GRCh38
rs759470362 Chr.9: 132828083-132828083 on Build GRCh38
rs370949050 Chr.9: 132828043-132828043 on Build GRCh38
rs183787418 Chr.9: 132828028-132828028 on Build GRCh38
rs1049083305 Chr.9: 132828132-132828132 on Build GRCh38
rs201332957 Chr.9: 132827988-132827988 on Build GRCh38
rs866414004 Chr.9: 132828052-132828052 on Build GRCh38
rs181800261 Chr.9: 132827993-132827993 on Build GRCh38
rs753802659 Chr.9: 132827992-132827992 on Build GRCh38
rs772965072 Chr.9: 132828062-132828062 on Build GRCh38
rs768761114 Chr.9: 132828054-132828054 on Build GRCh38
rs757916882 Chr.9: 132828002-132828002 on Build GRCh38
rs776137373 Chr.9: 132828050-132828050 on Build GRCh38
rs896694731 Chr.9: 132828026-132828026 on Build GRCh38
rs908959981 Chr.9: 132828117-132828117 on Build GRCh38
rs530817390 Chr.9: 132828146-132828146 on Build GRCh38
rs866957865 Chr.9: 132828122-132828122 on Build GRCh38
rs200457695 Chr.9: 132828001-132828001 on Build GRCh38
rs764290020 Chr.9: 132827974-132827974 on Build GRCh38
rs1036025983 Chr.9: 132827938-132827938 on Build GRCh38
rs751196477 Chr.9: 132828128-132828128 on Build GRCh38
rs776352135 Chr.9: 132828073-132828073 on Build GRCh38
rs763623170 Chr.9: 132828125-132828125 on Build GRCh38
rs921949194 Chr.9: 132827969-132827969 on Build GRCh38
rs933545217 Chr.9: 132827982-132827982 on Build GRCh38
rs114347984 Chr.9: 132828063-132828063 on Build GRCh38
rs541646315 Chr.9: 132828004-132828004 on Build GRCh38
rs767203480 Chr.9: 132828032-132828032 on Build GRCh38
rs912451628 Chr.9: 132827949-132827949 on Build GRCh38
rs563299354 Chr.9: 132828105-132828105 on Build GRCh38
rs375135938 Chr.9: 132828071-132828071 on Build GRCh38
rs751795457 Chr.9: 132827977-132827977 on Build GRCh38
rs148618920 Chr.9: 132828088-132828088 on Build GRCh38
rs942630753 Chr.9: 132827964-132827964 on Build GRCh38
rs963899945 Chr.9: 132828087-132828087 on Build GRCh38
rs749386096 Chr.9: 132828053-132828053 on Build GRCh38
rs768146247 Chr.9: 132827995-132828023 on Build GRCh38
rs765221586 Chr.9: 132828085-132828085 on Build GRCh38
rs902128647 Chr.9: 132827965-132827965 on Build GRCh38
rs574252711 Chr.9: 132828003-132828003 on Build GRCh38

View Full Product Details