Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10484765-10485028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD3
Entrez Gene ID
272
Gene Name
adenosine monophosphate deaminase 3
Gene Aliases
-
UniGene
Hs.501890
Gene Chromosome Location
Chr.11: 10450321-10507579 on Build GRCh38
Transcripts
NM_000480.2 NM_001025389.1 NM_001025390.1 NM_001172430.1 NM_001172431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2109248 c.774C>T p.P258P Chr.11: 10484977-10484977 on Build GRCh38
5654185 c.696C>T p.H232H Chr.11: 10484899-10484899 on Build GRCh38
3442882 c.714C>T p.L238L Chr.11: 10484917-10484917 on Build GRCh38
3442883 c.775G>A p.D259N Chr.11: 10484978-10484978 on Build GRCh38
77258 c.707G>T p.G236V Chr.11: 10484910-10484910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767419200 Chr.11: 10484900-10484900 on Build GRCh38
rs777969100 Chr.11: 10484968-10484968 on Build GRCh38
rs779934775 Chr.11: 10484949-10484949 on Build GRCh38
rs763586352 Chr.11: 10484872-10484872 on Build GRCh38
rs560405331 Chr.11: 10484943-10484943 on Build GRCh38
rs540688833 Chr.11: 10484942-10484942 on Build GRCh38
rs866866033 Chr.11: 10484927-10484927 on Build GRCh38
rs753498120 Chr.11: 10484795-10484795 on Build GRCh38
rs778226052 Chr.11: 10484797-10484797 on Build GRCh38
rs756810031 Chr.11: 10484796-10484796 on Build GRCh38
rs151198885 Chr.11: 10484821-10484821 on Build GRCh38
rs761302527 Chr.11: 10484904-10484904 on Build GRCh38
rs761582303 Chr.11: 10484996-10484996 on Build GRCh38
rs997715829 Chr.11: 10484816-10484816 on Build GRCh38
rs773101414 Chr.11: 10484864-10484864 on Build GRCh38
rs780910038 Chr.11: 10484959-10484959 on Build GRCh38
rs771555030 Chr.11: 10484877-10484877 on Build GRCh38
rs201491748 Chr.11: 10484788-10484788 on Build GRCh38
rs779879761 Chr.11: 10484827-10484827 on Build GRCh38
rs558311739 Chr.11: 10484863-10484863 on Build GRCh38
rs775710024 Chr.11: 10484995-10484995 on Build GRCh38
rs146416222 Chr.11: 10485002-10485002 on Build GRCh38
rs764866612 Chr.11: 10484905-10484905 on Build GRCh38
rs768905813 Chr.11: 10484841-10484841 on Build GRCh38
rs774908569 Chr.11: 10484893-10484893 on Build GRCh38
rs754789645 Chr.11: 10484958-10484958 on Build GRCh38
rs751459310 Chr.11: 10484954-10484954 on Build GRCh38
rs774305514 Chr.11: 10484894-10484894 on Build GRCh38
rs145610067 Chr.11: 10484978-10484978 on Build GRCh38
rs267602678 Chr.11: 10484976-10484976 on Build GRCh38
rs34319136 Chr.11: 10484899-10484899 on Build GRCh38
rs781424869 Chr.11: 10484844-10484844 on Build GRCh38
rs749309648 Chr.11: 10484973-10484973 on Build GRCh38
rs763820706 Chr.11: 10484789-10484789 on Build GRCh38
rs760057989 Chr.11: 10484980-10484980 on Build GRCh38
rs368122845 Chr.11: 10484965-10484965 on Build GRCh38
rs748269293 Chr.11: 10484857-10484857 on Build GRCh38
rs987924771 Chr.11: 10484932-10484932 on Build GRCh38
rs753551414 Chr.11: 10484901-10484901 on Build GRCh38
rs199564481 Chr.11: 10484999-10484999 on Build GRCh38
rs1803101 Chr.11: 10484843-10484843 on Build GRCh38
rs772567174 Chr.11: 10484994-10484994 on Build GRCh38
rs758409888 Chr.11: 10484946-10484946 on Build GRCh38
rs200373616 Chr.11: 10484830-10484830 on Build GRCh38
rs750375616 Chr.11: 10484798-10484798 on Build GRCh38
rs771609974 Chr.11: 10484977-10484977 on Build GRCh38
rs769907259 Chr.11: 10484966-10484966 on Build GRCh38
rs772392471 Chr.11: 10484954-10484955 on Build GRCh38
rs897571103 Chr.11: 10484809-10484809 on Build GRCh38
rs376653944 Chr.11: 10484802-10484802 on Build GRCh38
rs1050776692 Chr.11: 10484845-10484845 on Build GRCh38

View Full Product Details