Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151086733-151086982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
UniGene
Hs.647075
Gene Chromosome Location
Chr.7: 151085831-151144436 on Build GRCh38
Transcripts
NM_001042535.3 NM_001281300.1 NM_001308304.1 NM_031946.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5957673 c.89T>G p.L30R Chr.7: 151086830-151086830 on Build GRCh38
1673683 c.178C>T p.Q60* Chr.7: 151086919-151086919 on Build GRCh38
5957697 c.83A>G p.Q28R Chr.7: 151086824-151086824 on Build GRCh38
1622568 c.63G>C p.G21G Chr.7: 151086804-151086804 on Build GRCh38
5957688 c.94_95insGGGG p.C32fs*99 Chr.7: 151086835-151086836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138322898 Chr.7: 151086941-151086941 on Build GRCh38
rs746411238 Chr.7: 151086876-151086876 on Build GRCh38
rs750348682 Chr.7: 151086913-151086913 on Build GRCh38
rs750063767 Chr.7: 151086826-151086827 on Build GRCh38
rs773884793 Chr.7: 151086911-151086911 on Build GRCh38
rs766984834 Chr.7: 151086835-151086836 on Build GRCh38
rs777147483 Chr.7: 151086870-151086870 on Build GRCh38
rs572361798 Chr.7: 151086819-151086819 on Build GRCh38
rs749816387 Chr.7: 151086863-151086868 on Build GRCh38
rs753690096 Chr.7: 151086921-151086921 on Build GRCh38
rs776065932 Chr.7: 151086890-151086890 on Build GRCh38
rs555651682 Chr.7: 151086816-151086816 on Build GRCh38
rs770920056 Chr.7: 151086861-151086861 on Build GRCh38
rs761349408 Chr.7: 151086912-151086912 on Build GRCh38
rs867015348 Chr.7: 151086834-151086834 on Build GRCh38
rs552859803 Chr.7: 151086821-151086822 on Build GRCh38
rs1043076691 Chr.7: 151086859-151086864 on Build GRCh38
rs754779280 Chr.7: 151086925-151086925 on Build GRCh38
rs766154681 Chr.7: 151086915-151086915 on Build GRCh38
rs868413312 Chr.7: 151086788-151086788 on Build GRCh38
rs886043141 Chr.7: 151086821-151086821 on Build GRCh38
rs756868172 Chr.7: 151086815-151086835 on Build GRCh38
rs539642617 Chr.7: 151086813-151086814 on Build GRCh38
rs757337986 Chr.7: 151086943-151086943 on Build GRCh38
rs776100575 Chr.7: 151086833-151086835 on Build GRCh38
rs763783755 Chr.7: 151086910-151086910 on Build GRCh38
rs781169454 Chr.7: 151086952-151086952 on Build GRCh38
rs762964882 Chr.7: 151086806-151086835 on Build GRCh38
rs751333078 Chr.7: 151086815-151086816 on Build GRCh38
rs775200524 Chr.7: 151086830-151086830 on Build GRCh38
rs762684454 Chr.7: 151086909-151086909 on Build GRCh38
rs746383594 Chr.7: 151086954-151086954 on Build GRCh38
rs769408537 Chr.7: 151086824-151086824 on Build GRCh38
rs867838099 Chr.7: 151086835-151086835 on Build GRCh38
rs200151243 Chr.7: 151086914-151086914 on Build GRCh38
rs777904186 Chr.7: 151086936-151086936 on Build GRCh38

View Full Product Details