Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56361373-56361642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOF
Entrez Gene ID
319
Gene Name
apolipoprotein F
Gene Aliases
Apo-F, LTIP
UniGene
Hs.534302
Gene Chromosome Location
Chr.12: 56360571-56362799 on Build GRCh38
Transcripts
NM_001638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602431 c.643C>T p.R215* Chr.12: 56361563-56361563 on Build GRCh38
4555329 c.650G>A p.G217E Chr.12: 56361556-56361556 on Build GRCh38
941665 c.619C>A p.L207M Chr.12: 56361587-56361587 on Build GRCh38
4575595 c.705G>C p.G235G Chr.12: 56361501-56361501 on Build GRCh38
4662530 c.800A>G p.N267S Chr.12: 56361406-56361406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760810474 Chr.12: 56361480-56361480 on Build GRCh38
rs751178019 Chr.12: 56361548-56361549 on Build GRCh38
rs758079805 Chr.12: 56361495-56361495 on Build GRCh38
rs776818934 Chr.12: 56361571-56361571 on Build GRCh38
rs777019179 Chr.12: 56361486-56361487 on Build GRCh38
rs183126023 Chr.12: 56361609-56361609 on Build GRCh38
rs763784162 Chr.12: 56361598-56361598 on Build GRCh38
rs11575217 Chr.12: 56361516-56361516 on Build GRCh38
rs1001608340 Chr.12: 56361604-56361604 on Build GRCh38
rs373590737 Chr.12: 56361462-56361462 on Build GRCh38
rs778120152 Chr.12: 56361563-56361563 on Build GRCh38
rs749781824 Chr.12: 56361436-56361436 on Build GRCh38
rs778419868 Chr.12: 56361409-56361409 on Build GRCh38
rs746551111 Chr.12: 56361414-56361414 on Build GRCh38
rs762810631 Chr.12: 56361596-56361596 on Build GRCh38
rs766000132 Chr.12: 56361528-56361530 on Build GRCh38
rs771180959 Chr.12: 56361570-56361570 on Build GRCh38
rs774670066 Chr.12: 56361451-56361451 on Build GRCh38
rs755513463 Chr.12: 56361517-56361517 on Build GRCh38
rs762508541 Chr.12: 56361490-56361491 on Build GRCh38
rs780605685 Chr.12: 56361426-56361426 on Build GRCh38
rs779346057 Chr.12: 56361545-56361545 on Build GRCh38
rs374429340 Chr.12: 56361505-56361505 on Build GRCh38
rs769871876 Chr.12: 56361584-56361584 on Build GRCh38
rs771451803 Chr.12: 56361519-56361519 on Build GRCh38
rs754882582 Chr.12: 56361394-56361394 on Build GRCh38
rs1046744934 Chr.12: 56361498-56361498 on Build GRCh38
rs780660809 Chr.12: 56361506-56361506 on Build GRCh38
rs376665452 Chr.12: 56361494-56361494 on Build GRCh38
rs578174811 Chr.12: 56361487-56361487 on Build GRCh38
rs944280108 Chr.12: 56361508-56361508 on Build GRCh38
rs767118313 Chr.12: 56361615-56361615 on Build GRCh38
rs201865718 Chr.12: 56361420-56361420 on Build GRCh38
rs368571017 Chr.12: 56361510-56361510 on Build GRCh38
rs1047981635 Chr.12: 56361534-56361534 on Build GRCh38
rs765161945 Chr.12: 56361492-56361492 on Build GRCh38
rs999237166 Chr.12: 56361572-56361572 on Build GRCh38
rs754472787 Chr.12: 56361619-56361619 on Build GRCh38
rs769059248 Chr.12: 56361450-56361450 on Build GRCh38
rs759646761 Chr.12: 56361573-56361573 on Build GRCh38
rs375956731 Chr.12: 56361411-56361411 on Build GRCh38
rs371270374 Chr.12: 56361504-56361504 on Build GRCh38
rs748475007 Chr.12: 56361456-56361456 on Build GRCh38
rs554370648 Chr.12: 56361568-56361568 on Build GRCh38
rs1043565575 Chr.12: 56361565-56361565 on Build GRCh38
rs374949393 Chr.12: 56361591-56361591 on Build GRCh38
rs772261296 Chr.12: 56361562-56361562 on Build GRCh38
rs556656613 Chr.12: 56361486-56361486 on Build GRCh38
rs765108560 Chr.12: 56361401-56361401 on Build GRCh38
rs752443894 Chr.12: 56361403-56361403 on Build GRCh38
rs748562615 Chr.12: 56361551-56361551 on Build GRCh38
rs753758642 Chr.12: 56361393-56361393 on Build GRCh38
rs758037147 Chr.12: 56361410-56361410 on Build GRCh38
rs773253133 Chr.12: 56361474-56361474 on Build GRCh38
rs749960936 Chr.12: 56361618-56361618 on Build GRCh38
rs761343377 Chr.12: 56361614-56361614 on Build GRCh38
rs901546094 Chr.12: 56361478-56361478 on Build GRCh38
rs763820304 Chr.12: 56361503-56361503 on Build GRCh38

View Full Product Details