Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 106761147-106761408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABRA
Entrez Gene ID
137735
Gene Name
actin binding Rho activating protein
Gene Aliases
STARS
UniGene
Hs.374668
Gene Chromosome Location
Chr.8: 106759483-106770244 on Build GRCh38
Transcripts
NM_139166.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6591313 c.903T>C p.R301R Chr.8: 106761280-106761280 on Build GRCh38
5556470 c.839A>C p.H280P Chr.8: 106761344-106761344 on Build GRCh38
1094748 c.781_783delAAG p.K261delK Chr.8: 106761400-106761402 on Build GRCh38
3643843 c.875G>A p.G292E Chr.8: 106761308-106761308 on Build GRCh38
5746902 c.1036A>G p.K346E Chr.8: 106761147-106761147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182944429 Chr.8: 106761282-106761282 on Build GRCh38
rs781340771 Chr.8: 106761251-106761251 on Build GRCh38
rs978594063 Chr.8: 106761328-106761328 on Build GRCh38
rs767590580 Chr.8: 106761350-106761350 on Build GRCh38
rs760272225 Chr.8: 106761210-106761210 on Build GRCh38
rs753722518 Chr.8: 106761246-106761246 on Build GRCh38
rs750586739 Chr.8: 106761351-106761351 on Build GRCh38
rs751541398 Chr.8: 106761332-106761332 on Build GRCh38
rs755390954 Chr.8: 106761354-106761354 on Build GRCh38
rs372498827 Chr.8: 106761294-106761294 on Build GRCh38
rs781429118 Chr.8: 106761192-106761192 on Build GRCh38
rs746206328 Chr.8: 106761193-106761193 on Build GRCh38
rs200451420 Chr.8: 106761367-106761367 on Build GRCh38
rs780294176 Chr.8: 106761269-106761269 on Build GRCh38
rs759366681 Chr.8: 106761218-106761218 on Build GRCh38
rs749762231 Chr.8: 106761205-106761205 on Build GRCh38
rs777760983 Chr.8: 106761320-106761320 on Build GRCh38
rs374102076 Chr.8: 106761370-106761370 on Build GRCh38
rs756403095 Chr.8: 106761260-106761260 on Build GRCh38
rs780390590 Chr.8: 106761352-106761352 on Build GRCh38
rs779246467 Chr.8: 106761364-106761364 on Build GRCh38
rs745434102 Chr.8: 106761372-106761372 on Build GRCh38
rs757289786 Chr.8: 106761340-106761340 on Build GRCh38
rs1007502156 Chr.8: 106761323-106761323 on Build GRCh38
rs144057540 Chr.8: 106761281-106761281 on Build GRCh38
rs764154630 Chr.8: 106761242-106761242 on Build GRCh38
rs763919499 Chr.8: 106761326-106761326 on Build GRCh38
rs200193774 Chr.8: 106761321-106761321 on Build GRCh38
rs774972832 Chr.8: 106761284-106761284 on Build GRCh38
rs770163608 Chr.8: 106761195-106761195 on Build GRCh38
rs776447590 Chr.8: 106761215-106761215 on Build GRCh38
rs912722153 Chr.8: 106761289-106761289 on Build GRCh38
rs751558306 Chr.8: 106761243-106761243 on Build GRCh38
rs757373126 Chr.8: 106761249-106761249 on Build GRCh38
rs916365791 Chr.8: 106761185-106761185 on Build GRCh38
rs936678368 Chr.8: 106761386-106761386 on Build GRCh38
rs746470743 Chr.8: 106761285-106761285 on Build GRCh38
rs771134219 Chr.8: 106761184-106761184 on Build GRCh38
rs137891532 Chr.8: 106761258-106761258 on Build GRCh38
rs747203470 Chr.8: 106761359-106761359 on Build GRCh38
rs775933784 Chr.8: 106761196-106761196 on Build GRCh38
rs771552266 Chr.8: 106761206-106761206 on Build GRCh38
rs891290416 Chr.8: 106761312-106761312 on Build GRCh38
rs772583120 Chr.8: 106761209-106761209 on Build GRCh38
rs776357974 Chr.8: 106761319-106761319 on Build GRCh38
rs765131208 Chr.8: 106761219-106761219 on Build GRCh38
rs202190737 Chr.8: 106761353-106761353 on Build GRCh38
rs113347680 Chr.8: 106761347-106761347 on Build GRCh38
rs145316939 Chr.8: 106761237-106761237 on Build GRCh38
rs748668005 Chr.8: 106761366-106761366 on Build GRCh38
rs376042673 Chr.8: 106761227-106761227 on Build GRCh38
rs762851311 Chr.8: 106761325-106761325 on Build GRCh38

View Full Product Details