Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33014507-33014732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK2
Entrez Gene ID
204
Gene Name
adenylate kinase 2
Gene Aliases
ADK2
UniGene
Hs.470907
Gene Chromosome Location
Chr.1: 33007940-33036911 on Build GRCh38
Transcripts
NM_001199199.1 NM_001319139.1 NM_001319140.1 NM_001319141.1 NM_001319142.1 NM_001319143.1 NM_001625.3 NM_013411.4 NR_134976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134880 c.498+1G>A p.? Chr.1: 33014521-33014521 on Build GRCh38
7140837 c.489G>A p.M163I Chr.1: 33014531-33014531 on Build GRCh38
4860986 c.460G>T p.E154* Chr.1: 33014560-33014560 on Build GRCh38
228785 c.437C>T p.P146L Chr.1: 33014583-33014583 on Build GRCh38
4864211 c.459C>T p.H153H Chr.1: 33014561-33014561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749178848 Chr.1: 33014587-33014587 on Build GRCh38
rs773402998 Chr.1: 33014592-33014592 on Build GRCh38
rs371672441 Chr.1: 33014550-33014550 on Build GRCh38
rs779975688 Chr.1: 33014629-33014629 on Build GRCh38
rs11576688 Chr.1: 33014643-33014643 on Build GRCh38
rs150498169 Chr.1: 33014625-33014625 on Build GRCh38
rs142517578 Chr.1: 33014591-33014591 on Build GRCh38
rs1004153991 Chr.1: 33014690-33014690 on Build GRCh38
rs1037019280 Chr.1: 33014698-33014698 on Build GRCh38
rs572422227 Chr.1: 33014641-33014641 on Build GRCh38
rs758140036 Chr.1: 33014623-33014623 on Build GRCh38
rs771101990 Chr.1: 33014645-33014645 on Build GRCh38
rs56014743 Chr.1: 33014707-33014707 on Build GRCh38
rs756906982 Chr.1: 33014615-33014615 on Build GRCh38
rs148421308 Chr.1: 33014560-33014560 on Build GRCh38
rs768173996 Chr.1: 33014572-33014572 on Build GRCh38
rs987764285 Chr.1: 33014582-33014582 on Build GRCh38
rs773484833 Chr.1: 33014552-33014552 on Build GRCh38
rs371983494 Chr.1: 33014638-33014638 on Build GRCh38
rs760276418 Chr.1: 33014577-33014577 on Build GRCh38
rs745638583 Chr.1: 33014544-33014544 on Build GRCh38
rs774908387 Chr.1: 33014564-33014564 on Build GRCh38
rs777771065 Chr.1: 33014640-33014640 on Build GRCh38
rs200924521 Chr.1: 33014561-33014561 on Build GRCh38
rs541628571 Chr.1: 33014688-33014688 on Build GRCh38
rs770876157 Chr.1: 33014650-33014650 on Build GRCh38
rs367835240 Chr.1: 33014621-33014621 on Build GRCh38
rs149227118 Chr.1: 33014571-33014571 on Build GRCh38
rs751935223 Chr.1: 33014593-33014593 on Build GRCh38
rs764911232 Chr.1: 33014616-33014616 on Build GRCh38
rs552815574 Chr.1: 33014548-33014548 on Build GRCh38
rs751605899 Chr.1: 33014613-33014614 on Build GRCh38
rs754935209 Chr.1: 33014639-33014639 on Build GRCh38
rs146442876 Chr.1: 33014549-33014549 on Build GRCh38

View Full Product Details