Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61685904-61686131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf107
Entrez Gene ID
219621
Gene Name
chromosome 10 open reading frame 107
Gene Aliases
bA63A2.1
UniGene
Hs.673160
Gene Chromosome Location
Chr.10: 61662927-61766766 on Build GRCh38
Transcripts
NM_173554.2 XM_005269598.3 XM_005269599.4 XM_005269600.4 XM_006717690.3 XM_006717692.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603607 c.134C>T p.P45L Chr.10: 61686104-61686104 on Build GRCh38
5005424 c.144G>C p.S48S Chr.10: 61686114-61686114 on Build GRCh38
4562295 c.91G>A p.E31K Chr.10: 61686061-61686061 on Build GRCh38
369133 c.73C>G p.L25V Chr.10: 61686043-61686043 on Build GRCh38
1702539 c.143C>T p.S48L Chr.10: 61686113-61686113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893100968 Chr.10: 61685960-61685960 on Build GRCh38
rs750033930 Chr.10: 61685995-61685996 on Build GRCh38
rs367951018 Chr.10: 61686009-61686009 on Build GRCh38
rs761660716 Chr.10: 61685998-61685998 on Build GRCh38
rs1054477159 Chr.10: 61685934-61685934 on Build GRCh38
rs377014933 Chr.10: 61686045-61686045 on Build GRCh38
rs556574783 Chr.10: 61686023-61686023 on Build GRCh38
rs762857593 Chr.10: 61686101-61686101 on Build GRCh38
rs946341288 Chr.10: 61685962-61685962 on Build GRCh38
rs1016054175 Chr.10: 61686102-61686102 on Build GRCh38
rs761853158 Chr.10: 61686089-61686089 on Build GRCh38
rs772309016 Chr.10: 61686075-61686075 on Build GRCh38
rs12242793 Chr.10: 61685966-61685966 on Build GRCh38
rs749725890 Chr.10: 61686053-61686053 on Build GRCh38
rs200907159 Chr.10: 61686021-61686021 on Build GRCh38
rs754539555 Chr.10: 61686026-61686026 on Build GRCh38
rs1030902464 Chr.10: 61686055-61686055 on Build GRCh38
rs997791805 Chr.10: 61685987-61685987 on Build GRCh38
rs781637451 Chr.10: 61686028-61686028 on Build GRCh38
rs370292749 Chr.10: 61686068-61686068 on Build GRCh38
rs766097031 Chr.10: 61686022-61686022 on Build GRCh38
rs774678772 Chr.10: 61686066-61686066 on Build GRCh38
rs752059941 Chr.10: 61685985-61685985 on Build GRCh38
rs751257410 Chr.10: 61686109-61686109 on Build GRCh38
rs756415400 Chr.10: 61686047-61686047 on Build GRCh38
rs771026241 Chr.10: 61686054-61686054 on Build GRCh38
rs776643809 Chr.10: 61686080-61686080 on Build GRCh38
rs765126891 Chr.10: 61686000-61686000 on Build GRCh38
rs200469516 Chr.10: 61686043-61686043 on Build GRCh38
rs202247748 Chr.10: 61685993-61685993 on Build GRCh38
rs765899949 Chr.10: 61686107-61686107 on Build GRCh38
rs151048274 Chr.10: 61686091-61686091 on Build GRCh38
rs778100741 Chr.10: 61686049-61686049 on Build GRCh38
rs112711586 Chr.10: 61686108-61686108 on Build GRCh38
rs755725026 Chr.10: 61686062-61686062 on Build GRCh38
rs750033872 Chr.10: 61686018-61686018 on Build GRCh38
rs574991797 Chr.10: 61686096-61686096 on Build GRCh38

View Full Product Details