Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4626454-4626719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP3
Entrez Gene ID
10566
Gene Name
A-kinase anchoring protein 3
Gene Aliases
AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
UniGene
Hs.98397
Gene Chromosome Location
Chr.12: 4615508-4649047 on Build GRCh38
Transcripts
NM_001278309.1 NM_006422.3 XM_005253664.2 XM_011520908.1 XM_011520909.1 XM_011520910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2068246 c.2250A>C p.E750D Chr.12: 4626652-4626652 on Build GRCh38
5938307 c.2237G>A p.G746E Chr.12: 4626665-4626665 on Build GRCh38
5480094 c.2326G>A p.V776I Chr.12: 4626576-4626576 on Build GRCh38
6519893 c.2251G>T p.G751W Chr.12: 4626651-4626651 on Build GRCh38
2068245 c.2250A>C p.E750D Chr.12: 4626652-4626652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11063264 Chr.12: 4626607-4626607 on Build GRCh38
rs751392256 Chr.12: 4626655-4626655 on Build GRCh38
rs747685637 Chr.12: 4626590-4626590 on Build GRCh38
rs768650919 Chr.12: 4626577-4626577 on Build GRCh38
rs781228875 Chr.12: 4626666-4626666 on Build GRCh38
rs34781557 Chr.12: 4626634-4626634 on Build GRCh38
rs750328143 Chr.12: 4626483-4626484 on Build GRCh38
rs373282059 Chr.12: 4626643-4626643 on Build GRCh38
rs753969101 Chr.12: 4626533-4626533 on Build GRCh38
rs377722055 Chr.12: 4626487-4626487 on Build GRCh38
rs764687145 Chr.12: 4626633-4626633 on Build GRCh38
rs2072359 Chr.12: 4626568-4626568 on Build GRCh38
rs750217661 Chr.12: 4626564-4626564 on Build GRCh38
rs533468028 Chr.12: 4626516-4626516 on Build GRCh38
rs545624552 Chr.12: 4626532-4626532 on Build GRCh38
rs747637006 Chr.12: 4626669-4626669 on Build GRCh38
rs761032895 Chr.12: 4626631-4626631 on Build GRCh38
rs767890760 Chr.12: 4626545-4626545 on Build GRCh38
rs777399617 Chr.12: 4626677-4626677 on Build GRCh38
rs61757552 Chr.12: 4626522-4626522 on Build GRCh38
rs758694096 Chr.12: 4626648-4626648 on Build GRCh38
rs779316251 Chr.12: 4626547-4626547 on Build GRCh38
rs748873608 Chr.12: 4626679-4626679 on Build GRCh38
rs779141768 Chr.12: 4626649-4626649 on Build GRCh38
rs753202883 Chr.12: 4626510-4626510 on Build GRCh38
rs775931964 Chr.12: 4626620-4626620 on Build GRCh38
rs148740017 Chr.12: 4626576-4626576 on Build GRCh38
rs780518568 Chr.12: 4626589-4626589 on Build GRCh38
rs199800327 Chr.12: 4626608-4626608 on Build GRCh38
rs115289691 Chr.12: 4626635-4626635 on Build GRCh38
rs2072358 Chr.12: 4626571-4626571 on Build GRCh38
rs539568482 Chr.12: 4626671-4626671 on Build GRCh38
rs34392018 Chr.12: 4626604-4626604 on Build GRCh38
rs139674556 Chr.12: 4626660-4626660 on Build GRCh38
rs144371277 Chr.12: 4626521-4626521 on Build GRCh38

View Full Product Details