Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119895906-119896149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM30
Entrez Gene ID
11085
Gene Name
ADAM metallopeptidase domain 30
Gene Aliases
svph4
UniGene
Hs.283011
Gene Chromosome Location
Chr.1: 119893533-119896524 on Build GRCh38
Transcripts
NM_021794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057781 c.201G>T p.Q67H Chr.1: 119896136-119896136 on Build GRCh38
3788517 c.312T>A p.P104P Chr.1: 119896025-119896025 on Build GRCh38
5511563 c.198G>T p.L66L Chr.1: 119896139-119896139 on Build GRCh38
3740581 c.246T>A p.L82L Chr.1: 119896091-119896091 on Build GRCh38
4576181 c.345C>T p.S115S Chr.1: 119895992-119895992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017644291 Chr.1: 119896085-119896085 on Build GRCh38
rs899138662 Chr.1: 119895949-119895949 on Build GRCh38
rs767059190 Chr.1: 119895973-119895973 on Build GRCh38
rs776792195 Chr.1: 119896028-119896028 on Build GRCh38
rs190248331 Chr.1: 119895992-119895992 on Build GRCh38
rs140065466 Chr.1: 119895971-119895971 on Build GRCh38
rs758467993 Chr.1: 119896118-119896118 on Build GRCh38
rs375710052 Chr.1: 119895991-119895991 on Build GRCh38
rs1023282215 Chr.1: 119896000-119896000 on Build GRCh38
rs765610008 Chr.1: 119896030-119896030 on Build GRCh38
rs969046194 Chr.1: 119896034-119896034 on Build GRCh38
rs763487252 Chr.1: 119896052-119896052 on Build GRCh38
rs368265642 Chr.1: 119896029-119896029 on Build GRCh38
rs778622670 Chr.1: 119896093-119896093 on Build GRCh38
rs587685873 Chr.1: 119895999-119895999 on Build GRCh38
rs112598907 Chr.1: 119895943-119895943 on Build GRCh38
rs757098847 Chr.1: 119896090-119896090 on Build GRCh38
rs777612044 Chr.1: 119895983-119895984 on Build GRCh38
rs780974499 Chr.1: 119896100-119896100 on Build GRCh38
rs755709661 Chr.1: 119896072-119896072 on Build GRCh38
rs768935053 Chr.1: 119896026-119896026 on Build GRCh38
rs748090070 Chr.1: 119895941-119895942 on Build GRCh38
rs754502895 Chr.1: 119895972-119895972 on Build GRCh38
rs147294252 Chr.1: 119895954-119895954 on Build GRCh38
rs587666591 Chr.1: 119895990-119895990 on Build GRCh38
rs752310831 Chr.1: 119895976-119895976 on Build GRCh38
rs144460635 Chr.1: 119896016-119896016 on Build GRCh38
rs999369482 Chr.1: 119896061-119896061 on Build GRCh38
rs752265812 Chr.1: 119896071-119896071 on Build GRCh38
rs780223663 Chr.1: 119896124-119896124 on Build GRCh38
rs753539865 Chr.1: 119896089-119896089 on Build GRCh38
rs750354655 Chr.1: 119896102-119896102 on Build GRCh38
rs1037978740 Chr.1: 119895936-119895936 on Build GRCh38
rs369203475 Chr.1: 119895948-119895948 on Build GRCh38
rs966963262 Chr.1: 119896008-119896008 on Build GRCh38
rs763814394 Chr.1: 119896083-119896083 on Build GRCh38
rs777110616 Chr.1: 119895961-119895961 on Build GRCh38
rs767005904 Chr.1: 119896070-119896070 on Build GRCh38
rs750659972 Chr.1: 119896022-119896022 on Build GRCh38
rs146837536 Chr.1: 119895997-119895997 on Build GRCh38
rs376920145 Chr.1: 119895995-119895995 on Build GRCh38
rs762167444 Chr.1: 119895965-119895965 on Build GRCh38
rs756074989 Chr.1: 119895963-119895964 on Build GRCh38
rs149981088 Chr.1: 119895970-119895970 on Build GRCh38
rs371282723 Chr.1: 119896046-119896046 on Build GRCh38
rs773683718 Chr.1: 119896050-119896050 on Build GRCh38

View Full Product Details