Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 60059623-60059876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
UniGene
Hs.499725
Gene Chromosome Location
Chr.10: 60026298-60733526 on Build GRCh38
Transcripts
NM_001149.3 NM_001204403.1 NM_001204404.1 NM_001320874.1 NM_020987.4 XM_005269715.3 XM_006717796.3 XM_006717802.3 XM_011539708.2 XM_011539709.2 XM_017016110.1 XM_017016111.1 XM_017016112.1 XM_017016113.1 XM_017016114.1 XM_017016115.1 XM_017016116.1 XM_017016117.1 XM_017016118.1 XM_017016119.1 XM_017016120.1 XM_017016121.1 XM_017016122.1 XM_017016123.1 XM_017016124.1 XM_017016125.1 XM_017016126.1 XM_017016127.1 XM_017016128.1 XM_017016129.1 XM_017016130.1 XM_017016131.1 XM_017016132.1 XM_017016133.1 XM_017016134.1 XM_017016135.1 XM_017016136.1 XM_017016137.1 XM_017016138.1 XM_017016139.1 XM_017016140.1 XM_017016141.1 XM_017016142.1 XM_017016143.1 XM_017016144.1 XM_017016145.1 XM_017016146.1 XM_017016147.1 XM_017016148.1 XM_017016149.1 XM_017016150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
919214 c.2445C>T p.S815S Chr.10: 60059720-60059720 on Build GRCh38
216338 c.2451G>C p.L817F Chr.10: 60059714-60059714 on Build GRCh38
2148491 c.2427A>G p.A809A Chr.10: 60059738-60059738 on Build GRCh38
3978680 c.2357C>A p.T786N Chr.10: 60059808-60059808 on Build GRCh38
919215 c.2352C>T p.V784V Chr.10: 60059813-60059813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753340977 Chr.10: 60059816-60059816 on Build GRCh38
rs369435203 Chr.10: 60059825-60059825 on Build GRCh38
rs770952279 Chr.10: 60059705-60059705 on Build GRCh38
rs200010804 Chr.10: 60059806-60059806 on Build GRCh38
rs558378990 Chr.10: 60059747-60059747 on Build GRCh38
rs748748302 Chr.10: 60059686-60059686 on Build GRCh38
rs774594179 Chr.10: 60059733-60059733 on Build GRCh38
rs755411580 Chr.10: 60059677-60059677 on Build GRCh38
rs1050893490 Chr.10: 60059697-60059697 on Build GRCh38
rs1016446403 Chr.10: 60059679-60059679 on Build GRCh38
rs1036909947 Chr.10: 60059796-60059796 on Build GRCh38
rs768661868 Chr.10: 60059729-60059750 on Build GRCh38
rs199831681 Chr.10: 60059716-60059716 on Build GRCh38
rs199526263 Chr.10: 60059830-60059830 on Build GRCh38
rs746640430 Chr.10: 60059656-60059656 on Build GRCh38
rs201194312 Chr.10: 60059841-60059841 on Build GRCh38
rs561659383 Chr.10: 60059672-60059672 on Build GRCh38
rs754573454 Chr.10: 60059821-60059821 on Build GRCh38
rs765115990 Chr.10: 60059802-60059802 on Build GRCh38
rs993655279 Chr.10: 60059774-60059774 on Build GRCh38
rs770221013 Chr.10: 60059689-60059689 on Build GRCh38
rs151278674 Chr.10: 60059807-60059807 on Build GRCh38
rs564012648 Chr.10: 60059804-60059804 on Build GRCh38
rs749782714 Chr.10: 60059683-60059683 on Build GRCh38
rs749403750 Chr.10: 60059704-60059704 on Build GRCh38
rs751324021 Chr.10: 60059813-60059813 on Build GRCh38
rs12356776 Chr.10: 60059798-60059798 on Build GRCh38
rs898064106 Chr.10: 60059684-60059684 on Build GRCh38
rs373049128 Chr.10: 60059849-60059849 on Build GRCh38
rs1006896729 Chr.10: 60059674-60059674 on Build GRCh38
rs774840096 Chr.10: 60059714-60059714 on Build GRCh38
rs866697632 Chr.10: 60059831-60059831 on Build GRCh38
rs34390946 Chr.10: 60059791-60059792 on Build GRCh38
rs779873761 Chr.10: 60059649-60059649 on Build GRCh38
rs139092048 Chr.10: 60059785-60059785 on Build GRCh38
rs765957789 Chr.10: 60059812-60059812 on Build GRCh38
rs759670115 Chr.10: 60059734-60059734 on Build GRCh38
rs773646711 Chr.10: 60059693-60059693 on Build GRCh38
rs774298829 Chr.10: 60059847-60059847 on Build GRCh38
rs756463710 Chr.10: 60059839-60059839 on Build GRCh38

View Full Product Details