Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77694679-77694952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH1
Entrez Gene ID
8846
Gene Name
alkB homolog 1, histone H2A dioxygenase
Gene Aliases
ABH, ABH1, ALKBH, alkB, hABH
UniGene
Hs.94542
Gene Chromosome Location
Chr.14: 77672404-77708038 on Build GRCh38
Transcripts
NM_006020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333966 c.409A>G p.K137E Chr.14: 77694784-77694784 on Build GRCh38
5897074 c.293-5T>A p.? Chr.14: 77694905-77694905 on Build GRCh38
3769518 c.450C>T p.F150F Chr.14: 77694743-77694743 on Build GRCh38
6141193 c.417G>C p.E139D Chr.14: 77694776-77694776 on Build GRCh38
1323371 c.302T>G p.F101C Chr.14: 77694891-77694891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750218235 Chr.14: 77694782-77694782 on Build GRCh38
rs372795735 Chr.14: 77694742-77694742 on Build GRCh38
rs771136828 Chr.14: 77694855-77694855 on Build GRCh38
rs772373246 Chr.14: 77694716-77694716 on Build GRCh38
rs756625126 Chr.14: 77694718-77694718 on Build GRCh38
rs369190213 Chr.14: 77694748-77694748 on Build GRCh38
rs773595345 Chr.14: 77694720-77694720 on Build GRCh38
rs759182860 Chr.14: 77694732-77694732 on Build GRCh38
rs780327045 Chr.14: 77694791-77694791 on Build GRCh38
rs765368531 Chr.14: 77694927-77694927 on Build GRCh38
rs775301653 Chr.14: 77694858-77694858 on Build GRCh38
rs754110196 Chr.14: 77694911-77694911 on Build GRCh38
rs774711784 Chr.14: 77694926-77694928 on Build GRCh38
rs575190579 Chr.14: 77694875-77694875 on Build GRCh38
rs376909666 Chr.14: 77694859-77694859 on Build GRCh38
rs150673526 Chr.14: 77694821-77694821 on Build GRCh38
rs748761389 Chr.14: 77694920-77694921 on Build GRCh38
rs1000088072 Chr.14: 77694797-77694797 on Build GRCh38
rs1031208379 Chr.14: 77694811-77694811 on Build GRCh38
rs777432181 Chr.14: 77694777-77694779 on Build GRCh38
rs17825440 Chr.14: 77694788-77694788 on Build GRCh38
rs201927341 Chr.14: 77694739-77694739 on Build GRCh38
rs748695569 Chr.14: 77694817-77694817 on Build GRCh38
rs374787102 Chr.14: 77694743-77694743 on Build GRCh38
rs770851789 Chr.14: 77694796-77694796 on Build GRCh38
rs139082552 Chr.14: 77694864-77694864 on Build GRCh38
rs544107800 Chr.14: 77694909-77694909 on Build GRCh38
rs371958188 Chr.14: 77694776-77694776 on Build GRCh38
rs1002889537 Chr.14: 77694884-77694884 on Build GRCh38
rs558326346 Chr.14: 77694843-77694843 on Build GRCh38
rs142904199 Chr.14: 77694816-77694816 on Build GRCh38
rs772954642 Chr.14: 77694881-77694881 on Build GRCh38
rs767277460 Chr.14: 77694766-77694766 on Build GRCh38
rs761713671 Chr.14: 77694874-77694874 on Build GRCh38
rs754018189 Chr.14: 77694794-77694794 on Build GRCh38
rs755074590 Chr.14: 77694813-77694813 on Build GRCh38
rs1012305127 Chr.14: 77694745-77694745 on Build GRCh38
rs778062189 Chr.14: 77694838-77694838 on Build GRCh38
rs768319022 Chr.14: 77694860-77694860 on Build GRCh38
rs867557172 Chr.14: 77694846-77694846 on Build GRCh38
rs772628922 Chr.14: 77694829-77694829 on Build GRCh38
rs770473897 Chr.14: 77694921-77694921 on Build GRCh38
rs769478498 Chr.14: 77694738-77694738 on Build GRCh38
rs374257735 Chr.14: 77694886-77694886 on Build GRCh38
rs755162441 Chr.14: 77694913-77694913 on Build GRCh38
rs952017259 Chr.14: 77694916-77694916 on Build GRCh38
rs1013169197 Chr.14: 77694800-77694800 on Build GRCh38
rs760159598 Chr.14: 77694930-77694931 on Build GRCh38

View Full Product Details