Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25013471-25013722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
UniGene
Hs.529353
Gene Chromosome Location
Chr.20: 25006230-25058182 on Build GRCh38
Transcripts
NM_001252675.1 NM_001252676.1 NM_001252677.1 NM_032501.3 XM_006723659.1 XM_011529388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863273 c.1547G>T p.R516L Chr.20: 25013568-25013568 on Build GRCh38
4810118 c.1330+1G>C p.? Chr.20: 25013535-25013535 on Build GRCh38
5693614 c.1509G>T p.W503C Chr.20: 25013606-25013606 on Build GRCh38
3544847 c.1326C>T p.Y442Y Chr.20: 25013540-25013540 on Build GRCh38
5641529 c.1265G>T p.G422V Chr.20: 25013601-25013601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6115001 Chr.20: 25013639-25013639 on Build GRCh38
rs369571996 Chr.20: 25013674-25013674 on Build GRCh38
rs747031189 Chr.20: 25013568-25013568 on Build GRCh38
rs559882451 Chr.20: 25013680-25013680 on Build GRCh38
rs772815187 Chr.20: 25013686-25013686 on Build GRCh38
rs151321075 Chr.20: 25013654-25013654 on Build GRCh38
rs761161575 Chr.20: 25013618-25013618 on Build GRCh38
rs763565033 Chr.20: 25013509-25013509 on Build GRCh38
rs769662671 Chr.20: 25013497-25013497 on Build GRCh38
rs562522968 Chr.20: 25013671-25013671 on Build GRCh38
rs145378751 Chr.20: 25013579-25013579 on Build GRCh38
rs759044380 Chr.20: 25013563-25013563 on Build GRCh38
rs775357421 Chr.20: 25013504-25013504 on Build GRCh38
rs573299374 Chr.20: 25013681-25013681 on Build GRCh38
rs768637821 Chr.20: 25013603-25013603 on Build GRCh38
rs770653361 Chr.20: 25013694-25013694 on Build GRCh38
rs6050250 Chr.20: 25013657-25013657 on Build GRCh38
rs773916676 Chr.20: 25013510-25013510 on Build GRCh38
rs537842321 Chr.20: 25013651-25013651 on Build GRCh38
rs6050249 Chr.20: 25013653-25013653 on Build GRCh38
rs775305942 Chr.20: 25013587-25013587 on Build GRCh38
rs143222563 Chr.20: 25013573-25013573 on Build GRCh38
rs372826998 Chr.20: 25013529-25013529 on Build GRCh38
rs774284268 Chr.20: 25013604-25013604 on Build GRCh38
rs762928773 Chr.20: 25013505-25013505 on Build GRCh38
rs755254924 Chr.20: 25013546-25013546 on Build GRCh38
rs767082332 Chr.20: 25013622-25013622 on Build GRCh38
rs201013770 Chr.20: 25013565-25013565 on Build GRCh38
rs367670992 Chr.20: 25013638-25013638 on Build GRCh38
rs202113568 Chr.20: 25013688-25013688 on Build GRCh38
rs781232536 Chr.20: 25013569-25013569 on Build GRCh38
rs375259329 Chr.20: 25013660-25013660 on Build GRCh38
rs987551874 Chr.20: 25013676-25013676 on Build GRCh38
rs750095173 Chr.20: 25013540-25013540 on Build GRCh38
rs767019781 Chr.20: 25013533-25013533 on Build GRCh38
rs749217855 Chr.20: 25013588-25013588 on Build GRCh38
rs111277869 Chr.20: 25013656-25013656 on Build GRCh38
rs765663942 Chr.20: 25013634-25013634 on Build GRCh38
rs376439383 Chr.20: 25013630-25013630 on Build GRCh38
rs911967088 Chr.20: 25013677-25013677 on Build GRCh38
rs913248148 Chr.20: 25013652-25013652 on Build GRCh38
rs139870683 Chr.20: 25013558-25013558 on Build GRCh38
rs980305461 Chr.20: 25013615-25013615 on Build GRCh38
rs765652374 Chr.20: 25013557-25013557 on Build GRCh38
rs558530666 Chr.20: 25013633-25013633 on Build GRCh38
rs6050251 Chr.20: 25013697-25013697 on Build GRCh38

View Full Product Details