Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134942348-134942621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
UniGene
Hs.486520
Gene Chromosome Location
Chr.6: 134917390-134950122 on Build GRCh38
Transcripts
NM_001193480.1 NM_022568.3 NM_170771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366301 c.403G>T p.G135C Chr.6: 134942448-134942448 on Build GRCh38
5975789 c.313A>G p.M105V Chr.6: 134942538-134942538 on Build GRCh38
7412098 c.356C>T p.S119F Chr.6: 134942495-134942495 on Build GRCh38
5869358 c.374C>T p.T125M Chr.6: 134942477-134942477 on Build GRCh38
3620871 c.322C>T p.P108S Chr.6: 134942529-134942529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770780451 Chr.6: 134942382-134942382 on Build GRCh38
rs533886052 Chr.6: 134942432-134942432 on Build GRCh38
rs769410176 Chr.6: 134942597-134942597 on Build GRCh38
rs768733652 Chr.6: 134942566-134942566 on Build GRCh38
rs781744419 Chr.6: 134942427-134942427 on Build GRCh38
rs201195775 Chr.6: 134942410-134942410 on Build GRCh38
rs919371834 Chr.6: 134942528-134942528 on Build GRCh38
rs193096768 Chr.6: 134942591-134942591 on Build GRCh38
rs77178086 Chr.6: 134942464-134942464 on Build GRCh38
rs777827977 Chr.6: 134942435-134942435 on Build GRCh38
rs143707042 Chr.6: 134942448-134942448 on Build GRCh38
rs962815665 Chr.6: 134942490-134942490 on Build GRCh38
rs1016629869 Chr.6: 134942477-134942477 on Build GRCh38
rs145368914 Chr.6: 134942538-134942538 on Build GRCh38
rs750074237 Chr.6: 134942505-134942505 on Build GRCh38
rs960957758 Chr.6: 134942563-134942563 on Build GRCh38
rs776291395 Chr.6: 134942494-134942494 on Build GRCh38
rs375965488 Chr.6: 134942441-134942441 on Build GRCh38
rs753123197 Chr.6: 134942431-134942431 on Build GRCh38
rs189584943 Chr.6: 134942512-134942512 on Build GRCh38
rs746063668 Chr.6: 134942547-134942547 on Build GRCh38
rs753313800 Chr.6: 134942524-134942524 on Build GRCh38
rs778196657 Chr.6: 134942526-134942526 on Build GRCh38
rs755263036 Chr.6: 134942426-134942426 on Build GRCh38
rs1028168544 Chr.6: 134942434-134942434 on Build GRCh38
rs532277046 Chr.6: 134942379-134942379 on Build GRCh38
rs144258828 Chr.6: 134942465-134942465 on Build GRCh38
rs771965268 Chr.6: 134942551-134942551 on Build GRCh38
rs143751098 Chr.6: 134942525-134942525 on Build GRCh38
rs372728487 Chr.6: 134942409-134942409 on Build GRCh38
rs752384284 Chr.6: 134942444-134942444 on Build GRCh38
rs755526654 Chr.6: 134942377-134942377 on Build GRCh38
rs771717393 Chr.6: 134942465-134942481 on Build GRCh38
rs776674323 Chr.6: 134942570-134942570 on Build GRCh38
rs775259675 Chr.6: 134942401-134942401 on Build GRCh38
rs140344994 Chr.6: 134942420-134942420 on Build GRCh38
rs200230491 Chr.6: 134942415-134942415 on Build GRCh38
rs201838947 Chr.6: 134942442-134942442 on Build GRCh38
rs747990200 Chr.6: 134942388-134942389 on Build GRCh38
rs753948428 Chr.6: 134942533-134942533 on Build GRCh38
rs149214929 Chr.6: 134942553-134942553 on Build GRCh38
rs369000455 Chr.6: 134942378-134942378 on Build GRCh38
rs369657445 Chr.6: 134942476-134942476 on Build GRCh38
rs749501426 Chr.6: 134942380-134942380 on Build GRCh38
rs867338689 Chr.6: 134942522-134942522 on Build GRCh38
rs773461025 Chr.6: 134942511-134942514 on Build GRCh38
rs746757712 Chr.6: 134942558-134942558 on Build GRCh38
rs757293946 Chr.6: 134942537-134942537 on Build GRCh38
rs140050752 Chr.6: 134942500-134942500 on Build GRCh38
rs775551985 Chr.6: 134942480-134942480 on Build GRCh38
rs761362230 Chr.6: 134942499-134942499 on Build GRCh38
rs3813342 Chr.6: 134942419-134942419 on Build GRCh38
rs759276112 Chr.6: 134942391-134942391 on Build GRCh38
rs150041987 Chr.6: 134942502-134942502 on Build GRCh38
rs760522133 Chr.6: 134942492-134942492 on Build GRCh38
rs147800650 Chr.6: 134942493-134942493 on Build GRCh38
rs541243826 Chr.6: 134942425-134942425 on Build GRCh38
rs767519872 Chr.6: 134942396-134942396 on Build GRCh38
rs767761456 Chr.6: 134942519-134942519 on Build GRCh38
rs568940837 Chr.6: 134942403-134942403 on Build GRCh38
rs202009000 Chr.6: 134942388-134942388 on Build GRCh38

View Full Product Details