Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6415665-6415924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHSRP
Entrez Gene ID
8570
Gene Name
KH-type splicing regulatory protein
Gene Aliases
FBP2, FUBP2, KSRP
UniGene
Hs.727344
Gene Chromosome Location
Chr.19: 6413104-6424811 on Build GRCh38
Transcripts
NM_003685.2 XM_005259668.4 XM_011528395.2 XM_017027408.1 XM_017027409.1 XM_017027410.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389488 c.1753C>T p.Q585* Chr.19: 6415669-6415669 on Build GRCh38
215802 c.1640G>A p.W547* Chr.19: 6415855-6415855 on Build GRCh38
6040790 c.1724C>T p.A575V Chr.19: 6415698-6415698 on Build GRCh38
3738136 c.1599-6C>G p.? Chr.19: 6415902-6415902 on Build GRCh38
5218234 c.1608_1609insG p.P537fs*25 Chr.19: 6415886-6415887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62107578 Chr.19: 6415700-6415700 on Build GRCh38
rs1001574119 Chr.19: 6415719-6415719 on Build GRCh38
rs775434772 Chr.19: 6415892-6415892 on Build GRCh38
rs1034003521 Chr.19: 6415715-6415715 on Build GRCh38
rs753736667 Chr.19: 6415902-6415902 on Build GRCh38
rs770374295 Chr.19: 6415741-6415741 on Build GRCh38
rs756355398 Chr.19: 6415698-6415698 on Build GRCh38
rs1045667557 Chr.19: 6415864-6415864 on Build GRCh38
rs761172391 Chr.19: 6415765-6415765 on Build GRCh38
rs772039285 Chr.19: 6415789-6415789 on Build GRCh38
rs145698454 Chr.19: 6415724-6415724 on Build GRCh38
rs780747211 Chr.19: 6415827-6415827 on Build GRCh38
rs762935980 Chr.19: 6415781-6415781 on Build GRCh38
rs745443610 Chr.19: 6415801-6415801 on Build GRCh38
rs765244740 Chr.19: 6415778-6415778 on Build GRCh38
rs779028251 Chr.19: 6415760-6415760 on Build GRCh38
rs975746514 Chr.19: 6415774-6415774 on Build GRCh38
rs746368203 Chr.19: 6415751-6415751 on Build GRCh38
rs748936288 Chr.19: 6415733-6415733 on Build GRCh38
rs202196104 Chr.19: 6415776-6415776 on Build GRCh38
rs769111737 Chr.19: 6415775-6415775 on Build GRCh38
rs374342583 Chr.19: 6415787-6415787 on Build GRCh38
rs755939315 Chr.19: 6415786-6415786 on Build GRCh38
rs949333804 Chr.19: 6415863-6415863 on Build GRCh38
rs756869408 Chr.19: 6415791-6415791 on Build GRCh38
rs763139301 Chr.19: 6415872-6415872 on Build GRCh38
rs8103311 Chr.19: 6415784-6415784 on Build GRCh38
rs763740814 Chr.19: 6415887-6415887 on Build GRCh38
rs780969352 Chr.19: 6415702-6415702 on Build GRCh38
rs897766667 Chr.19: 6415703-6415703 on Build GRCh38
rs992428014 Chr.19: 6415723-6415723 on Build GRCh38
rs377190857 Chr.19: 6415759-6415759 on Build GRCh38
rs951199113 Chr.19: 6415737-6415737 on Build GRCh38
rs897710422 Chr.19: 6415828-6415828 on Build GRCh38
rs751474510 Chr.19: 6415697-6415697 on Build GRCh38
rs368528290 Chr.19: 6415836-6415836 on Build GRCh38
rs778428488 Chr.19: 6415794-6415794 on Build GRCh38
rs868450284 Chr.19: 6415855-6415855 on Build GRCh38
rs554522870 Chr.19: 6415866-6415866 on Build GRCh38
rs777077352 Chr.19: 6415841-6415841 on Build GRCh38
rs760119287 Chr.19: 6415784-6415784 on Build GRCh38
rs191726896 Chr.19: 6415718-6415718 on Build GRCh38
rs375302000 Chr.19: 6415712-6415712 on Build GRCh38
rs777216403 Chr.19: 6415780-6415781 on Build GRCh38
rs931683084 Chr.19: 6415785-6415785 on Build GRCh38
rs1022202220 Chr.19: 6415693-6415693 on Build GRCh38
rs766993994 Chr.19: 6415759-6415760 on Build GRCh38
rs568677654 Chr.19: 6415826-6415826 on Build GRCh38
rs907209539 Chr.19: 6415865-6415865 on Build GRCh38
rs1039656607 Chr.19: 6415753-6415753 on Build GRCh38
rs778432931 Chr.19: 6415750-6415750 on Build GRCh38
rs891836238 Chr.19: 6415689-6415689 on Build GRCh38
rs73565847 Chr.19: 6415893-6415893 on Build GRCh38
rs750448941 Chr.19: 6415780-6415780 on Build GRCh38
rs749403979 Chr.19: 6415708-6415708 on Build GRCh38
rs765320420 Chr.19: 6415691-6415691 on Build GRCh38
rs537771953 Chr.19: 6415845-6415845 on Build GRCh38
rs766410802 Chr.19: 6415887-6415887 on Build GRCh38
rs762045989 Chr.19: 6415777-6415777 on Build GRCh38

View Full Product Details