Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 125318480-125318751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT4
Entrez Gene ID
79633
Gene Name
FAT atypical cadherin 4
Gene Aliases
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2
UniGene
Hs.563205
Gene Chromosome Location
Chr.4: 125314913-125492932 on Build GRCh38
Transcripts
NM_001291285.1 NM_001291303.1 NM_024582.4 XM_011532236.2 XM_011532237.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4478026 c.2203C>T p.R735W Chr.4: 125318614-125318614 on Build GRCh38
4121933 c.2116A>G p.S706G Chr.4: 125318527-125318527 on Build GRCh38
732705 c.2180G>A p.S727N Chr.4: 125318591-125318591 on Build GRCh38
732704 c.2180G>A p.S727N Chr.4: 125318591-125318591 on Build GRCh38
4478027 c.2203C>T p.R735W Chr.4: 125318614-125318614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763316688 Chr.4: 125318581-125318581 on Build GRCh38
rs755138266 Chr.4: 125318659-125318659 on Build GRCh38
rs994174271 Chr.4: 125318649-125318649 on Build GRCh38
rs950629016 Chr.4: 125318690-125318690 on Build GRCh38
rs763643835 Chr.4: 125318620-125318620 on Build GRCh38
rs377230037 Chr.4: 125318676-125318676 on Build GRCh38
rs780265799 Chr.4: 125318516-125318516 on Build GRCh38
rs753314680 Chr.4: 125318625-125318625 on Build GRCh38
rs150906264 Chr.4: 125318529-125318529 on Build GRCh38
rs201172737 Chr.4: 125318636-125318636 on Build GRCh38
rs200036436 Chr.4: 125318677-125318677 on Build GRCh38
rs530759297 Chr.4: 125318597-125318597 on Build GRCh38
rs1003773324 Chr.4: 125318571-125318571 on Build GRCh38
rs1007361222 Chr.4: 125318563-125318563 on Build GRCh38
rs758608981 Chr.4: 125318501-125318501 on Build GRCh38
rs781771280 Chr.4: 125318537-125318537 on Build GRCh38
rs751758773 Chr.4: 125318658-125318658 on Build GRCh38
rs570502199 Chr.4: 125318684-125318684 on Build GRCh38
rs200550937 Chr.4: 125318720-125318720 on Build GRCh38
rs187427286 Chr.4: 125318727-125318727 on Build GRCh38
rs758147582 Chr.4: 125318640-125318640 on Build GRCh38
rs774554680 Chr.4: 125318698-125318698 on Build GRCh38
rs1019279953 Chr.4: 125318699-125318699 on Build GRCh38
rs780174313 Chr.4: 125318650-125318650 on Build GRCh38
rs1012999985 Chr.4: 125318626-125318626 on Build GRCh38
rs946876507 Chr.4: 125318511-125318511 on Build GRCh38
rs200512777 Chr.4: 125318627-125318627 on Build GRCh38
rs775165068 Chr.4: 125318614-125318614 on Build GRCh38
rs866969841 Chr.4: 125318575-125318575 on Build GRCh38
rs367762836 Chr.4: 125318569-125318569 on Build GRCh38
rs372605427 Chr.4: 125318671-125318671 on Build GRCh38
rs748717152 Chr.4: 125318556-125318556 on Build GRCh38
rs902754252 Chr.4: 125318533-125318533 on Build GRCh38
rs747065140 Chr.4: 125318527-125318527 on Build GRCh38
rs746714727 Chr.4: 125318701-125318701 on Build GRCh38
rs771276464 Chr.4: 125318697-125318697 on Build GRCh38
rs770396155 Chr.4: 125318561-125318561 on Build GRCh38
rs771623506 Chr.4: 125318593-125318593 on Build GRCh38
rs376468352 Chr.4: 125318615-125318615 on Build GRCh38
rs749643997 Chr.4: 125318694-125318694 on Build GRCh38
rs1042560643 Chr.4: 125318514-125318514 on Build GRCh38

View Full Product Details