Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14851210-14851437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
UniGene
Hs.534169
Gene Chromosome Location
Chr.10: 14838160-14871741 on Build GRCh38
Transcripts
NM_016299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149721 c.529C>T p.L177F Chr.10: 14851280-14851280 on Build GRCh38
916601 c.482C>A p.A161D Chr.10: 14851233-14851233 on Build GRCh38
7224483 c.565G>T p.G189* Chr.10: 14851316-14851316 on Build GRCh38
6208601 c.518C>T p.P173L Chr.10: 14851269-14851269 on Build GRCh38
3358694 c.562A>T p.T188S Chr.10: 14851313-14851313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766100141 Chr.10: 14851268-14851268 on Build GRCh38
rs757800139 Chr.10: 14851291-14851291 on Build GRCh38
rs1010658254 Chr.10: 14851302-14851302 on Build GRCh38
rs1021594295 Chr.10: 14851309-14851309 on Build GRCh38
rs753596081 Chr.10: 14851269-14851269 on Build GRCh38
rs112799123 Chr.10: 14851377-14851377 on Build GRCh38
rs980178060 Chr.10: 14851349-14851349 on Build GRCh38
rs746230499 Chr.10: 14851306-14851306 on Build GRCh38
rs764835486 Chr.10: 14851279-14851279 on Build GRCh38
rs570039143 Chr.10: 14851253-14851253 on Build GRCh38
rs569279090 Chr.10: 14851236-14851236 on Build GRCh38
rs146276897 Chr.10: 14851286-14851286 on Build GRCh38
rs780197521 Chr.10: 14851336-14851336 on Build GRCh38
rs960180250 Chr.10: 14851390-14851390 on Build GRCh38
rs531326507 Chr.10: 14851270-14851270 on Build GRCh38
rs760611940 Chr.10: 14851267-14851267 on Build GRCh38
rs918853186 Chr.10: 14851406-14851406 on Build GRCh38
rs749540348 Chr.10: 14851350-14851350 on Build GRCh38
rs868767840 Chr.10: 14851289-14851289 on Build GRCh38
rs762380702 Chr.10: 14851393-14851393 on Build GRCh38
rs538274140 Chr.10: 14851254-14851254 on Build GRCh38
rs756517663 Chr.10: 14851314-14851314 on Build GRCh38
rs773045153 Chr.10: 14851265-14851265 on Build GRCh38
rs761810912 Chr.10: 14851264-14851264 on Build GRCh38
rs1034539039 Chr.10: 14851356-14851356 on Build GRCh38
rs969243456 Chr.10: 14851313-14851313 on Build GRCh38
rs781611125 Chr.10: 14851292-14851292 on Build GRCh38

View Full Product Details