Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24551345-24551618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319
Entrez Gene ID
9856
Gene Name
KIAA0319
Gene Aliases
DYLX2, DYX2, NMIG
UniGene
Hs.26441
Gene Chromosome Location
Chr.6: 24540131-24646211 on Build GRCh38
Transcripts
NM_001168374.1 NM_001168375.1 NM_001168376.1 NM_001168377.1 NM_001252328.1 NM_014809.3 XM_011515025.2 XM_011515026.2 XM_017011541.1 XM_017011542.1 XM_017011543.1 XM_017011544.1 XM_017011545.1 XM_017011546.1 XM_017011547.1 XM_017011548.1 XM_017011549.1 XM_017011550.1 XM_017011551.1 XM_017011552.1 XM_017011553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6892194 c.2958G>A p.R986R Chr.6: 24551516-24551516 on Build GRCh38
6693105 c.2977_2978insA p.T993fs*8 Chr.6: 24551496-24551497 on Build GRCh38
5404905 c.2995G>A p.D999N Chr.6: 24551479-24551479 on Build GRCh38
3921156 c.2970G>A p.R990R Chr.6: 24551504-24551504 on Build GRCh38
6892195 c.2931G>A p.R977R Chr.6: 24551516-24551516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772648728 Chr.6: 24551591-24551591 on Build GRCh38
rs745535230 Chr.6: 24551449-24551449 on Build GRCh38
rs910328530 Chr.6: 24551583-24551583 on Build GRCh38
rs866430826 Chr.6: 24551452-24551452 on Build GRCh38
rs755365931 Chr.6: 24551556-24551556 on Build GRCh38
rs755557530 Chr.6: 24551511-24551511 on Build GRCh38
rs758762480 Chr.6: 24551567-24551567 on Build GRCh38
rs993765622 Chr.6: 24551485-24551485 on Build GRCh38
rs186070494 Chr.6: 24551443-24551443 on Build GRCh38
rs370847933 Chr.6: 24551538-24551538 on Build GRCh38
rs766663941 Chr.6: 24551546-24551546 on Build GRCh38
rs767679949 Chr.6: 24551395-24551395 on Build GRCh38
rs375671437 Chr.6: 24551564-24551564 on Build GRCh38
rs779340540 Chr.6: 24551559-24551559 on Build GRCh38
rs138705676 Chr.6: 24551456-24551456 on Build GRCh38
rs753910096 Chr.6: 24551517-24551517 on Build GRCh38
rs775285717 Chr.6: 24551458-24551458 on Build GRCh38
rs373654218 Chr.6: 24551555-24551555 on Build GRCh38
rs572954657 Chr.6: 24551557-24551557 on Build GRCh38
rs749607427 Chr.6: 24551408-24551408 on Build GRCh38
rs807534 Chr.6: 24551436-24551436 on Build GRCh38
rs779526013 Chr.6: 24551414-24551414 on Build GRCh38
rs807535 Chr.6: 24551501-24551501 on Build GRCh38
rs374547514 Chr.6: 24551390-24551390 on Build GRCh38
rs754286941 Chr.6: 24551507-24551507 on Build GRCh38
rs748684777 Chr.6: 24551581-24551588 on Build GRCh38
rs755569182 Chr.6: 24551410-24551410 on Build GRCh38
rs756504723 Chr.6: 24551397-24551397 on Build GRCh38
rs199600231 Chr.6: 24551404-24551404 on Build GRCh38
rs984975236 Chr.6: 24551514-24551514 on Build GRCh38
rs780762682 Chr.6: 24551576-24551576 on Build GRCh38
rs764057053 Chr.6: 24551470-24551470 on Build GRCh38
rs2076313 Chr.6: 24551396-24551396 on Build GRCh38
rs868593953 Chr.6: 24551512-24551512 on Build GRCh38
rs763787738 Chr.6: 24551376-24551376 on Build GRCh38
rs757327624 Chr.6: 24551393-24551393 on Build GRCh38
rs746543791 Chr.6: 24551435-24551435 on Build GRCh38
rs765486492 Chr.6: 24551590-24551590 on Build GRCh38
rs542002661 Chr.6: 24551526-24551526 on Build GRCh38
rs377548888 Chr.6: 24551459-24551459 on Build GRCh38
rs755051590 Chr.6: 24551549-24551549 on Build GRCh38
rs866642138 Chr.6: 24551483-24551483 on Build GRCh38

View Full Product Details