Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144511092-144511335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD3
Entrez Gene ID
113655
Gene Name
major facilitator superfamily domain containing 3
Gene Aliases
-
UniGene
Hs.7678
Gene Chromosome Location
Chr.8: 144508081-144511228 on Build GRCh38
Transcripts
NM_138431.2 NR_130120.1 XM_011516806.2 XM_017013005.1 NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_138431.2 NR_130120.1 XM_011516806.2 XM_017013005.1 NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198783 c.1200C>T p.P400P Chr.8: 144511125-144511125 on Build GRCh38
5027011 c.1182C>T p.L394L Chr.8: 144511107-144511107 on Build GRCh38
6712013 c.1197T>C p.F399F Chr.8: 144511122-144511122 on Build GRCh38
4410840 c.1222C>T p.P408S Chr.8: 144511147-144511147 on Build GRCh38
3834299 c.1234C>G p.L412V Chr.8: 144511159-144511159 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756831197 Chr.8: 144511180-144511180 on Build GRCh38
rs777372134 Chr.8: 144511178-144511178 on Build GRCh38
rs374406640 Chr.8: 144511173-144511173 on Build GRCh38
rs747652501 Chr.8: 144511114-144511114 on Build GRCh38
rs758036287 Chr.8: 144511115-144511115 on Build GRCh38
rs1007688755 Chr.8: 144511161-144511161 on Build GRCh38
rs773309344 Chr.8: 144511140-144511140 on Build GRCh38
rs866356147 Chr.8: 144511204-144511204 on Build GRCh38
rs1022008379 Chr.8: 144511282-144511282 on Build GRCh38
rs556947725 Chr.8: 144511283-144511283 on Build GRCh38
rs1016971780 Chr.8: 144511269-144511269 on Build GRCh38
rs368438583 Chr.8: 144511201-144511201 on Build GRCh38
rs373704217 Chr.8: 144511205-144511205 on Build GRCh38
rs780966733 Chr.8: 144511182-144511182 on Build GRCh38
rs759007988 Chr.8: 144511177-144511177 on Build GRCh38
rs542293319 Chr.8: 144511126-144511126 on Build GRCh38
rs770642610 Chr.8: 144511130-144511130 on Build GRCh38
rs763220003 Chr.8: 144511191-144511191 on Build GRCh38
rs988016143 Chr.8: 144511295-144511296 on Build GRCh38
rs776248905 Chr.8: 144511217-144511217 on Build GRCh38
rs745717348 Chr.8: 144511134-144511134 on Build GRCh38
rs139004523 Chr.8: 144511138-144511138 on Build GRCh38
rs998813713 Chr.8: 144511197-144511197 on Build GRCh38
rs779511959 Chr.8: 144511113-144511113 on Build GRCh38
rs769616407 Chr.8: 144511135-144511135 on Build GRCh38
rs17850833 Chr.8: 144511162-144511162 on Build GRCh38
rs777431156 Chr.8: 144511119-144511119 on Build GRCh38
rs745637170 Chr.8: 144511184-144511184 on Build GRCh38
rs368010480 Chr.8: 144511179-144511179 on Build GRCh38
rs1043462052 Chr.8: 144511145-144511145 on Build GRCh38
rs751671277 Chr.8: 144511223-144511223 on Build GRCh38
rs760847213 Chr.8: 144511209-144511209 on Build GRCh38
rs771051109 Chr.8: 144511210-144511210 on Build GRCh38
rs959111004 Chr.8: 144511262-144511262 on Build GRCh38
rs1037793012 Chr.8: 144511163-144511163 on Build GRCh38
rs776270377 Chr.8: 144511132-144511132 on Build GRCh38
rs775465367 Chr.8: 144511136-144511136 on Build GRCh38
rs370663265 Chr.8: 144511144-144511144 on Build GRCh38
rs113964706 Chr.8: 144511281-144511281 on Build GRCh38
rs1033339194 Chr.8: 144511224-144511224 on Build GRCh38
rs866822564 Chr.8: 144511286-144511286 on Build GRCh38
rs575210383 Chr.8: 144511294-144511294 on Build GRCh38
rs143762437 Chr.8: 144511156-144511156 on Build GRCh38
rs751118468 Chr.8: 144511220-144511221 on Build GRCh38
rs769574636 Chr.8: 144511187-144511187 on Build GRCh38
rs762891711 Chr.8: 144511137-144511137 on Build GRCh38
rs753192223 Chr.8: 144511174-144511174 on Build GRCh38
rs977635246 Chr.8: 144511287-144511287 on Build GRCh38
rs766975589 Chr.8: 144511196-144511197 on Build GRCh38
rs535744209 Chr.8: 144511241-144511241 on Build GRCh38
rs372265598 Chr.8: 144511215-144511215 on Build GRCh38
rs377594353 Chr.8: 144511295-144511295 on Build GRCh38
rs754117686 Chr.8: 144511148-144511148 on Build GRCh38
rs768601470 Chr.8: 144511192-144511192 on Build GRCh38
rs993359779 Chr.8: 144511193-144511193 on Build GRCh38
rs545465885 Chr.8: 144511310-144511310 on Build GRCh38
rs746624115 Chr.8: 144511125-144511125 on Build GRCh38
rs771712122 Chr.8: 144511158-144511159 on Build GRCh38
rs112193719 Chr.8: 144511234-144511234 on Build GRCh38
rs760758309 Chr.8: 144511141-144511141 on Build GRCh38
rs552740699 Chr.8: 144511212-144511212 on Build GRCh38
rs984206661 Chr.8: 144511238-144511238 on Build GRCh38
rs368092848 Chr.8: 144511116-144511116 on Build GRCh38

View Full Product Details