Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66638821-66639094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954187 c.524G>A p.R175Q Chr.7: 66638886-66638886 on Build GRCh38
75097 c.530G>A p.R177H Chr.7: 66638892-66638892 on Build GRCh38
5976455 c.626C>T p.S209F Chr.7: 66638988-66638988 on Build GRCh38
6692116 c.593C>T p.P198L Chr.7: 66638955-66638955 on Build GRCh38
747279 c.618C>G p.L206L Chr.7: 66638980-66638980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745960603 Chr.7: 66638966-66638966 on Build GRCh38
rs745917176 Chr.7: 66638868-66638868 on Build GRCh38
rs765770848 Chr.7: 66638991-66638991 on Build GRCh38
rs369862253 Chr.7: 66639027-66639027 on Build GRCh38
rs768234025 Chr.7: 66638849-66638849 on Build GRCh38
rs773060529 Chr.7: 66638896-66638896 on Build GRCh38
rs770883670 Chr.7: 66638962-66638962 on Build GRCh38
rs201990302 Chr.7: 66639061-66639061 on Build GRCh38
rs749192199 Chr.7: 66638956-66638956 on Build GRCh38
rs754365629 Chr.7: 66638984-66638984 on Build GRCh38
rs774660202 Chr.7: 66638871-66638871 on Build GRCh38
rs777890772 Chr.7: 66638928-66638928 on Build GRCh38
rs372120833 Chr.7: 66638968-66638968 on Build GRCh38
rs755711171 Chr.7: 66639002-66639002 on Build GRCh38
rs200907176 Chr.7: 66638879-66638879 on Build GRCh38
rs762356641 Chr.7: 66638985-66638985 on Build GRCh38
rs727502785 Chr.7: 66638956-66638956 on Build GRCh38
rs752360120 Chr.7: 66638998-66638998 on Build GRCh38
rs368001837 Chr.7: 66638895-66638895 on Build GRCh38
rs747676224 Chr.7: 66638913-66638913 on Build GRCh38
rs376332394 Chr.7: 66638994-66638994 on Build GRCh38
rs772238582 Chr.7: 66638974-66638974 on Build GRCh38
rs117194263 Chr.7: 66639016-66639016 on Build GRCh38
rs776046806 Chr.7: 66638852-66638852 on Build GRCh38
rs376944331 Chr.7: 66638983-66638983 on Build GRCh38
rs796052689 Chr.7: 66639066-66639066 on Build GRCh38
rs773227811 Chr.7: 66638867-66638867 on Build GRCh38
rs372150992 Chr.7: 66639049-66639049 on Build GRCh38
rs769365291 Chr.7: 66638926-66638926 on Build GRCh38
rs387907246 Chr.7: 66638912-66638912 on Build GRCh38
rs267601551 Chr.7: 66638864-66638864 on Build GRCh38
rs754654827 Chr.7: 66638904-66638904 on Build GRCh38
rs930655842 Chr.7: 66638980-66638980 on Build GRCh38
rs796052688 Chr.7: 66638898-66638905 on Build GRCh38
rs199965432 Chr.7: 66638982-66638982 on Build GRCh38
rs142379946 Chr.7: 66638869-66638869 on Build GRCh38
rs769912440 Chr.7: 66638860-66638860 on Build GRCh38
rs750811871 Chr.7: 66638993-66638993 on Build GRCh38
rs767744597 Chr.7: 66638884-66638884 on Build GRCh38
rs746601430 Chr.7: 66638845-66638845 on Build GRCh38
rs866499042 Chr.7: 66638881-66638881 on Build GRCh38
rs758709078 Chr.7: 66638886-66638886 on Build GRCh38
rs774050685 Chr.7: 66638964-66638964 on Build GRCh38
rs568539835 Chr.7: 66638885-66638885 on Build GRCh38
rs140600524 Chr.7: 66638978-66638978 on Build GRCh38
rs747689300 Chr.7: 66638856-66638856 on Build GRCh38
rs151338966 Chr.7: 66638976-66638976 on Build GRCh38
rs966874768 Chr.7: 66638873-66638873 on Build GRCh38
rs750770298 Chr.7: 66638903-66638903 on Build GRCh38

View Full Product Details