Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23415144-23415395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6719473 c.5409C>T p.I1803I Chr.14: 23415145-23415145 on Build GRCh38
416236 c.5323A>G p.T1775A Chr.14: 23415231-23415231 on Build GRCh38
1629249 c.5317C>T p.Q1773* Chr.14: 23415237-23415237 on Build GRCh38
6719471 c.5374G>A p.D1792N Chr.14: 23415180-23415180 on Build GRCh38
954739 c.5287G>A p.A1763T Chr.14: 23415267-23415267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369429415 Chr.14: 23415336-23415336 on Build GRCh38
rs774733006 Chr.14: 23415202-23415202 on Build GRCh38
rs876657886 Chr.14: 23415251-23415252 on Build GRCh38
rs786204383 Chr.14: 23415357-23415357 on Build GRCh38
rs771626451 Chr.14: 23415361-23415361 on Build GRCh38
rs397516241 Chr.14: 23415252-23415252 on Build GRCh38
rs771432461 Chr.14: 23415268-23415268 on Build GRCh38
rs377182974 Chr.14: 23415344-23415344 on Build GRCh38
rs763412364 Chr.14: 23415191-23415191 on Build GRCh38
rs758213465 Chr.14: 23415238-23415238 on Build GRCh38
rs139222507 Chr.14: 23415249-23415249 on Build GRCh38
rs780938189 Chr.14: 23415318-23415318 on Build GRCh38
rs772148835 Chr.14: 23415374-23415374 on Build GRCh38
rs773795845 Chr.14: 23415266-23415266 on Build GRCh38
rs759628313 Chr.14: 23415206-23415206 on Build GRCh38
rs554943023 Chr.14: 23415296-23415296 on Build GRCh38
rs397516247 Chr.14: 23415174-23415174 on Build GRCh38
rs760841010 Chr.14: 23415297-23415297 on Build GRCh38
rs797044597 Chr.14: 23415175-23415177 on Build GRCh38
rs773427105 Chr.14: 23415185-23415185 on Build GRCh38
rs730880815 Chr.14: 23415237-23415237 on Build GRCh38
rs397516246 Chr.14: 23415212-23415212 on Build GRCh38
rs200939753 Chr.14: 23415225-23415225 on Build GRCh38
rs780553495 Chr.14: 23415251-23415251 on Build GRCh38
rs727504385 Chr.14: 23415210-23415210 on Build GRCh38
rs746667903 Chr.14: 23415244-23415244 on Build GRCh38
rs752105000 Chr.14: 23415333-23415333 on Build GRCh38
rs200205856 Chr.14: 23415335-23415335 on Build GRCh38
rs121913654 Chr.14: 23415176-23415176 on Build GRCh38
rs201875538 Chr.14: 23415303-23415303 on Build GRCh38
rs267606909 Chr.14: 23415258-23415258 on Build GRCh38
rs876661179 Chr.14: 23415188-23415190 on Build GRCh38
rs369437262 Chr.14: 23415228-23415228 on Build GRCh38
rs750389662 Chr.14: 23415299-23415299 on Build GRCh38
rs727504355 Chr.14: 23415267-23415267 on Build GRCh38
rs587779396 Chr.14: 23415174-23415176 on Build GRCh38
rs748598020 Chr.14: 23415168-23415168 on Build GRCh38
rs766156556 Chr.14: 23415306-23415306 on Build GRCh38
rs765036184 Chr.14: 23415298-23415298 on Build GRCh38
rs752882625 Chr.14: 23415214-23415214 on Build GRCh38
rs770389433 Chr.14: 23415180-23415180 on Build GRCh38
rs397516244 Chr.14: 23415222-23415222 on Build GRCh38
rs730880935 Chr.14: 23415200-23415202 on Build GRCh38
rs201171029 Chr.14: 23415193-23415193 on Build GRCh38
rs765263589 Chr.14: 23415226-23415226 on Build GRCh38
rs532242410 Chr.14: 23415334-23415334 on Build GRCh38
rs544452304 Chr.14: 23415253-23415253 on Build GRCh38
rs397516245 Chr.14: 23415213-23415213 on Build GRCh38
rs770165807 Chr.14: 23415261-23415261 on Build GRCh38
rs1052787042 Chr.14: 23415366-23415366 on Build GRCh38
rs201089330 Chr.14: 23415356-23415356 on Build GRCh38
rs45468101 Chr.14: 23415315-23415315 on Build GRCh38
rs759821521 Chr.14: 23415273-23415273 on Build GRCh38
rs767904246 Chr.14: 23415287-23415287 on Build GRCh38
rs867017523 Chr.14: 23415190-23415190 on Build GRCh38
rs746278750 Chr.14: 23415363-23415363 on Build GRCh38
rs779625738 Chr.14: 23415241-23415241 on Build GRCh38
rs45582836 Chr.14: 23415362-23415362 on Build GRCh38

View Full Product Details