Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124454662-124454929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRD2
Entrez Gene ID
347088
Gene Name
adhesion G protein-coupled receptor D2
Gene Aliases
GPR144, PGR24
UniGene
Hs.454099
Gene Chromosome Location
Chr.9: 124451087-124477795 on Build GRCh38
Transcripts
XM_006717348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5791849 c.1176C>A p.D392E Chr.9: 124454854-124454854 on Build GRCh38
4654427 c.1249C>A p.L417M Chr.9: 124454927-124454927 on Build GRCh38
6341498 c.1191C>T p.V397V Chr.9: 124454869-124454869 on Build GRCh38
7369990 c.1217C>A p.A406D Chr.9: 124454895-124454895 on Build GRCh38
1208647 c.1211C>T p.A404V Chr.9: 124454889-124454889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578119316 Chr.9: 124454863-124454863 on Build GRCh38
rs1038313701 Chr.9: 124454704-124454704 on Build GRCh38
rs905461802 Chr.9: 124454896-124454896 on Build GRCh38
rs1002182007 Chr.9: 124454902-124454902 on Build GRCh38
rs779591406 Chr.9: 124454840-124454840 on Build GRCh38
rs938497757 Chr.9: 124454710-124454710 on Build GRCh38
rs949617142 Chr.9: 124454864-124454864 on Build GRCh38
rs780265344 Chr.9: 124454878-124454878 on Build GRCh38
rs932294227 Chr.9: 124454691-124454691 on Build GRCh38
rs995392922 Chr.9: 124454729-124454729 on Build GRCh38
rs180947607 Chr.9: 124454831-124454831 on Build GRCh38
rs867363318 Chr.9: 124454879-124454879 on Build GRCh38
rs1036889537 Chr.9: 124454713-124454713 on Build GRCh38
rs983766701 Chr.9: 124454724-124454724 on Build GRCh38
rs909606050 Chr.9: 124454793-124454793 on Build GRCh38
rs776960587 Chr.9: 124454877-124454877 on Build GRCh38
rs1048012017 Chr.9: 124454792-124454792 on Build GRCh38
rs897990069 Chr.9: 124454850-124454850 on Build GRCh38
rs747326349 Chr.9: 124454890-124454890 on Build GRCh38
rs1031853113 Chr.9: 124454893-124454893 on Build GRCh38
rs939758487 Chr.9: 124454824-124454824 on Build GRCh38
rs746307556 Chr.9: 124454851-124454851 on Build GRCh38
rs557119578 Chr.9: 124454693-124454693 on Build GRCh38

View Full Product Details