Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10291710-10291967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM5
Entrez Gene ID
7087
Gene Name
intercellular adhesion molecule 5
Gene Aliases
TLCN, TLN
UniGene
Hs.465862
Gene Chromosome Location
Chr.19: 10289352-10296778 on Build GRCh38
Transcripts
NM_003259.3 XM_011528229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177656 c.578G>C p.G193A Chr.19: 10291714-10291714 on Build GRCh38
3148990 c.588C>A p.F196L Chr.19: 10291724-10291724 on Build GRCh38
1641045 c.591G>A p.S197S Chr.19: 10291727-10291727 on Build GRCh38
4634351 c.670T>C p.F224L Chr.19: 10291806-10291806 on Build GRCh38
3756263 c.618G>T p.P206P Chr.19: 10291754-10291754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990878988 Chr.19: 10291936-10291936 on Build GRCh38
rs78617778 Chr.19: 10291935-10291935 on Build GRCh38
rs776400501 Chr.19: 10291765-10291765 on Build GRCh38
rs573759852 Chr.19: 10291853-10291853 on Build GRCh38
rs998067808 Chr.19: 10291753-10291753 on Build GRCh38
rs759899104 Chr.19: 10291828-10291828 on Build GRCh38
rs1016393766 Chr.19: 10291881-10291881 on Build GRCh38
rs200196487 Chr.19: 10291849-10291849 on Build GRCh38
rs115870389 Chr.19: 10291754-10291754 on Build GRCh38
rs779366879 Chr.19: 10291746-10291746 on Build GRCh38
rs534071602 Chr.19: 10291771-10291771 on Build GRCh38
rs78432167 Chr.19: 10291934-10291934 on Build GRCh38
rs181890650 Chr.19: 10291931-10291931 on Build GRCh38
rs768070421 Chr.19: 10291835-10291835 on Build GRCh38
rs771193899 Chr.19: 10291791-10291791 on Build GRCh38
rs35701141 Chr.19: 10291743-10291744 on Build GRCh38
rs917803134 Chr.19: 10291933-10291933 on Build GRCh38
rs774835810 Chr.19: 10291805-10291805 on Build GRCh38
rs754057362 Chr.19: 10291857-10291857 on Build GRCh38
rs909222112 Chr.19: 10291919-10291919 on Build GRCh38
rs377362542 Chr.19: 10291830-10291830 on Build GRCh38
rs953552277 Chr.19: 10291810-10291810 on Build GRCh38
rs752116580 Chr.19: 10291926-10291926 on Build GRCh38
rs962007570 Chr.19: 10291908-10291908 on Build GRCh38
rs764202759 Chr.19: 10291854-10291854 on Build GRCh38
rs566775151 Chr.19: 10291757-10291757 on Build GRCh38

View Full Product Details