Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92919879-92920150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6
Entrez Gene ID
54536
Gene Name
exocyst complex component 6
Gene Aliases
EXOC6A, SEC15, SEC15L, SEC15L1, SEC15L3, Sec15p
UniGene
Hs.655657
Gene Chromosome Location
Chr.10: 92826831-93059494 on Build GRCh38
Transcripts
NM_001013848.3 NM_001319194.1 NM_001319195.1 NM_001319200.1 NM_019053.5 XM_017016344.1 XM_017016345.1 XM_017016346.1 XM_017016347.1 XM_017016348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758940 c.895T>G p.F299V Chr.10: 92920009-92920009 on Build GRCh38
1969730 c.846T>C p.D282D Chr.10: 92920008-92920008 on Build GRCh38
1205826 c.850T>C p.S284P Chr.10: 92920012-92920012 on Build GRCh38
1969731 c.831T>C p.D277D Chr.10: 92920008-92920008 on Build GRCh38
1474866 c.874A>G p.I292V Chr.10: 92920036-92920036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371866338 Chr.10: 92919935-92919935 on Build GRCh38
rs758312862 Chr.10: 92919954-92919955 on Build GRCh38
rs151098804 Chr.10: 92919989-92919989 on Build GRCh38
rs752519779 Chr.10: 92920078-92920078 on Build GRCh38
rs777151619 Chr.10: 92920038-92920038 on Build GRCh38
rs981179295 Chr.10: 92919910-92919911 on Build GRCh38
rs778565649 Chr.10: 92920032-92920032 on Build GRCh38
rs563282365 Chr.10: 92920025-92920025 on Build GRCh38
rs770261543 Chr.10: 92920044-92920044 on Build GRCh38
rs773222023 Chr.10: 92920048-92920048 on Build GRCh38
rs748691647 Chr.10: 92920041-92920041 on Build GRCh38
rs931538818 Chr.10: 92920106-92920106 on Build GRCh38
rs763082662 Chr.10: 92920053-92920053 on Build GRCh38
rs760091184 Chr.10: 92920014-92920014 on Build GRCh38
rs774584497 Chr.10: 92920058-92920058 on Build GRCh38
rs751613711 Chr.10: 92919962-92919962 on Build GRCh38
rs932544767 Chr.10: 92920104-92920104 on Build GRCh38
rs757724300 Chr.10: 92919994-92919994 on Build GRCh38
rs777477654 Chr.10: 92920003-92920003 on Build GRCh38
rs756336522 Chr.10: 92919958-92919958 on Build GRCh38
rs748821778 Chr.10: 92920013-92920013 on Build GRCh38
rs201436972 Chr.10: 92919951-92919951 on Build GRCh38
rs747343247 Chr.10: 92920033-92920033 on Build GRCh38
rs992611762 Chr.10: 92919974-92919974 on Build GRCh38
rs1050480575 Chr.10: 92920113-92920113 on Build GRCh38
rs750482779 Chr.10: 92919950-92919950 on Build GRCh38
rs185657687 Chr.10: 92919906-92919906 on Build GRCh38
rs1008976965 Chr.10: 92920114-92920114 on Build GRCh38
rs767462180 Chr.10: 92920076-92920076 on Build GRCh38
rs143757847 Chr.10: 92919949-92919949 on Build GRCh38
rs529748973 Chr.10: 92919961-92919961 on Build GRCh38
rs768956810 Chr.10: 92920036-92920036 on Build GRCh38
rs1045357077 Chr.10: 92920059-92920059 on Build GRCh38
rs766514008 Chr.10: 92920054-92920054 on Build GRCh38
rs781706057 Chr.10: 92920107-92920107 on Build GRCh38
rs754744180 Chr.10: 92919973-92919973 on Build GRCh38
rs752644074 Chr.10: 92919991-92919991 on Build GRCh38
rs924476981 Chr.10: 92919929-92919929 on Build GRCh38
rs761900463 Chr.10: 92919931-92919931 on Build GRCh38
rs759436164 Chr.10: 92920064-92920064 on Build GRCh38

View Full Product Details