Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35577502-35577737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
UniGene
Hs.591190
Gene Chromosome Location
Chr.17: 35574795-35578637 on Build GRCh38
Transcripts
NM_000286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220216 c.184A>C p.I62L Chr.17: 35577534-35577534 on Build GRCh38
1324031 c.194T>C p.L65P Chr.17: 35577524-35577524 on Build GRCh38
7121867 c.205C>T p.L69L Chr.17: 35577513-35577513 on Build GRCh38
460364 c.178G>C p.D60H Chr.17: 35577540-35577540 on Build GRCh38
5649302 c.157_158insT p.G53fs*8 Chr.17: 35577560-35577561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771304850 Chr.17: 35577563-35577564 on Build GRCh38
rs766393993 Chr.17: 35577527-35577527 on Build GRCh38
rs774416658 Chr.17: 35577625-35577625 on Build GRCh38
rs779094557 Chr.17: 35577593-35577602 on Build GRCh38
rs968031925 Chr.17: 35577670-35577670 on Build GRCh38
rs758005394 Chr.17: 35577552-35577552 on Build GRCh38
rs373145272 Chr.17: 35577562-35577562 on Build GRCh38
rs775665571 Chr.17: 35577611-35577611 on Build GRCh38
rs979347663 Chr.17: 35577671-35577671 on Build GRCh38
rs746311261 Chr.17: 35577591-35577591 on Build GRCh38
rs760582512 Chr.17: 35577626-35577626 on Build GRCh38
rs956219162 Chr.17: 35577653-35577653 on Build GRCh38
rs756156620 Chr.17: 35577563-35577563 on Build GRCh38
rs1054829779 Chr.17: 35577580-35577580 on Build GRCh38
rs770008536 Chr.17: 35577603-35577603 on Build GRCh38
rs369640430 Chr.17: 35577529-35577529 on Build GRCh38
rs375578252 Chr.17: 35577677-35577677 on Build GRCh38
rs766018784 Chr.17: 35577630-35577630 on Build GRCh38
rs753823640 Chr.17: 35577632-35577632 on Build GRCh38
rs777258588 Chr.17: 35577553-35577553 on Build GRCh38
rs763301937 Chr.17: 35577617-35577617 on Build GRCh38
rs994372639 Chr.17: 35577657-35577657 on Build GRCh38
rs768965527 Chr.17: 35577624-35577624 on Build GRCh38
rs527389243 Chr.17: 35577705-35577705 on Build GRCh38
rs1008239545 Chr.17: 35577610-35577610 on Build GRCh38
rs988859283 Chr.17: 35577654-35577654 on Build GRCh38
rs923069079 Chr.17: 35577525-35577525 on Build GRCh38
rs754484547 Chr.17: 35577536-35577536 on Build GRCh38
rs36033015 Chr.17: 35577592-35577592 on Build GRCh38
rs926411530 Chr.17: 35577694-35577694 on Build GRCh38
rs936321326 Chr.17: 35577575-35577575 on Build GRCh38
rs777263696 Chr.17: 35577583-35577583 on Build GRCh38
rs764155994 Chr.17: 35577672-35577672 on Build GRCh38
rs913800657 Chr.17: 35577656-35577656 on Build GRCh38
rs897293816 Chr.17: 35577656-35577657 on Build GRCh38
rs771188005 Chr.17: 35577569-35577569 on Build GRCh38
rs759416939 Chr.17: 35577642-35577642 on Build GRCh38
rs747820399 Chr.17: 35577542-35577542 on Build GRCh38
rs778333662 Chr.17: 35577540-35577540 on Build GRCh38
rs143036724 Chr.17: 35577646-35577646 on Build GRCh38

View Full Product Details