Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5867271-5867504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT5
Entrez Gene ID
2527
Gene Name
fucosyltransferase 5
Gene Aliases
FUC-TV
UniGene
Hs.631843
Gene Chromosome Location
Chr.19: 5865826-5870540 on Build GRCh38
Transcripts
NM_002034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1481626 c.423G>T p.Q141H Chr.19: 5867303-5867303 on Build GRCh38
4685895 c.302C>T p.A101V Chr.19: 5867424-5867424 on Build GRCh38
6667894 c.390delC p.S131fs*117 Chr.19: 5867336-5867336 on Build GRCh38
6667895 c.339C>T p.Y113Y Chr.19: 5867387-5867387 on Build GRCh38
3540721 c.435G>A p.W145* Chr.19: 5867291-5867291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762157139 Chr.19: 5867339-5867339 on Build GRCh38
rs781355035 Chr.19: 5867312-5867312 on Build GRCh38
rs762401144 Chr.19: 5867294-5867294 on Build GRCh38
rs111394159 Chr.19: 5867335-5867335 on Build GRCh38
rs1013613076 Chr.19: 5867308-5867308 on Build GRCh38
rs61731655 Chr.19: 5867303-5867303 on Build GRCh38
rs148489400 Chr.19: 5867402-5867402 on Build GRCh38
rs771830771 Chr.19: 5867316-5867316 on Build GRCh38
rs770395527 Chr.19: 5867341-5867341 on Build GRCh38
rs199516299 Chr.19: 5867365-5867365 on Build GRCh38
rs566802540 Chr.19: 5867442-5867442 on Build GRCh38
rs140732692 Chr.19: 5867424-5867424 on Build GRCh38
rs139667433 Chr.19: 5867322-5867322 on Build GRCh38
rs759674318 Chr.19: 5867413-5867413 on Build GRCh38
rs780946375 Chr.19: 5867444-5867444 on Build GRCh38
rs779896589 Chr.19: 5867318-5867318 on Build GRCh38
rs758420902 Chr.19: 5867428-5867428 on Build GRCh38
rs200621532 Chr.19: 5867315-5867315 on Build GRCh38
rs760851573 Chr.19: 5867423-5867423 on Build GRCh38
rs868466988 Chr.19: 5867484-5867484 on Build GRCh38
rs536359180 Chr.19: 5867419-5867419 on Build GRCh38
rs1020181970 Chr.19: 5867403-5867403 on Build GRCh38
rs61731563 Chr.19: 5867448-5867448 on Build GRCh38
rs763481225 Chr.19: 5867401-5867401 on Build GRCh38
rs746693814 Chr.19: 5867321-5867321 on Build GRCh38
rs1018909398 Chr.19: 5867391-5867391 on Build GRCh38
rs201760120 Chr.19: 5867306-5867306 on Build GRCh38
rs1037057757 Chr.19: 5867357-5867357 on Build GRCh38
rs374135977 Chr.19: 5867480-5867480 on Build GRCh38
rs370088810 Chr.19: 5867309-5867309 on Build GRCh38
rs141356302 Chr.19: 5867461-5867461 on Build GRCh38
rs201253650 Chr.19: 5867382-5867382 on Build GRCh38
rs200650200 Chr.19: 5867452-5867452 on Build GRCh38
rs550781895 Chr.19: 5867395-5867395 on Build GRCh38
rs201728770 Chr.19: 5867412-5867412 on Build GRCh38
rs373593937 Chr.19: 5867429-5867429 on Build GRCh38
rs758230697 Chr.19: 5867387-5867387 on Build GRCh38
rs773748982 Chr.19: 5867398-5867398 on Build GRCh38
rs370745789 Chr.19: 5867468-5867468 on Build GRCh38
rs747750504 Chr.19: 5867397-5867397 on Build GRCh38
rs760833851 Chr.19: 5867483-5867483 on Build GRCh38
rs749122551 Chr.19: 5867313-5867313 on Build GRCh38
rs767999133 Chr.19: 5867324-5867324 on Build GRCh38
rs189234701 Chr.19: 5867458-5867458 on Build GRCh38
rs145907945 Chr.19: 5867425-5867425 on Build GRCh38
rs752890602 Chr.19: 5867374-5867374 on Build GRCh38
rs750215709 Chr.19: 5867386-5867386 on Build GRCh38
rs562121143 Chr.19: 5867296-5867296 on Build GRCh38
rs767430432 Chr.19: 5867373-5867373 on Build GRCh38
rs780013169 Chr.19: 5867393-5867393 on Build GRCh38
rs375231882 Chr.19: 5867307-5867307 on Build GRCh38
rs1037131192 Chr.19: 5867476-5867476 on Build GRCh38
rs757129901 Chr.19: 5867379-5867379 on Build GRCh38
rs773550928 Chr.19: 5867346-5867346 on Build GRCh38
rs144634186 Chr.19: 5867481-5867481 on Build GRCh38
rs556886147 Chr.19: 5867463-5867463 on Build GRCh38
rs765460174 Chr.19: 5867295-5867295 on Build GRCh38
rs776214188 Chr.19: 5867334-5867334 on Build GRCh38
rs757225388 Chr.19: 5867314-5867314 on Build GRCh38
rs149784295 Chr.19: 5867372-5867372 on Build GRCh38
rs373534405 Chr.19: 5867366-5867366 on Build GRCh38
rs145858323 Chr.19: 5867394-5867394 on Build GRCh38

View Full Product Details