Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137307057-137307292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD3
Entrez Gene ID
54932
Gene Name
exonuclease 3'-5' domain containing 3
Gene Aliases
mut-7
UniGene
Hs.495553
Gene Chromosome Location
Chr.9: 137306894-137423262 on Build GRCh38
Transcripts
NM_017820.4 XM_011518810.1 XM_011518814.1 XM_011518815.1 XM_017014861.1 XM_017014862.1 XM_017014863.1 XM_017014864.1 XM_017014865.1 XM_017014866.1 XM_017014867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657041 c.2523C>T p.D841D Chr.9: 137307058-137307058 on Build GRCh38
6380550 c.2389C>A p.L797M Chr.9: 137307192-137307192 on Build GRCh38
6223145 c.2480G>A p.R827Q Chr.9: 137307101-137307101 on Build GRCh38
4152197 c.2358C>A p.D786E Chr.9: 137307223-137307223 on Build GRCh38
1284789 c.2493C>T p.C831C Chr.9: 137307088-137307088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751950815 Chr.9: 137307249-137307249 on Build GRCh38
rs751355575 Chr.9: 137307241-137307242 on Build GRCh38
rs560190764 Chr.9: 137307164-137307164 on Build GRCh38
rs761568310 Chr.9: 137307198-137307198 on Build GRCh38
rs377043054 Chr.9: 137307118-137307118 on Build GRCh38
rs532420019 Chr.9: 137307184-137307184 on Build GRCh38
rs879717411 Chr.9: 137307084-137307084 on Build GRCh38
rs773524595 Chr.9: 137307141-137307141 on Build GRCh38
rs745429103 Chr.9: 137307105-137307105 on Build GRCh38
rs28685844 Chr.9: 137307152-137307152 on Build GRCh38
rs964109682 Chr.9: 137307252-137307252 on Build GRCh38
rs868715951 Chr.9: 137307239-137307239 on Build GRCh38
rs199943268 Chr.9: 137307162-137307162 on Build GRCh38
rs769764009 Chr.9: 137307115-137307115 on Build GRCh38
rs777360321 Chr.9: 137307094-137307094 on Build GRCh38
rs542445427 Chr.9: 137307132-137307132 on Build GRCh38
rs200971534 Chr.9: 137307219-137307219 on Build GRCh38
rs775321602 Chr.9: 137307141-137307141 on Build GRCh38
rs371442328 Chr.9: 137307130-137307130 on Build GRCh38
rs776184180 Chr.9: 137307102-137307102 on Build GRCh38
rs774262529 Chr.9: 137307188-137307188 on Build GRCh38
rs540192818 Chr.9: 137307159-137307159 on Build GRCh38
rs760916828 Chr.9: 137307146-137307146 on Build GRCh38
rs746694933 Chr.9: 137307098-137307098 on Build GRCh38
rs111790802 Chr.9: 137307151-137307151 on Build GRCh38
rs752815262 Chr.9: 137307218-137307218 on Build GRCh38
rs558615597 Chr.9: 137307163-137307163 on Build GRCh38
rs748448428 Chr.9: 137307089-137307089 on Build GRCh38
rs373543265 Chr.9: 137307082-137307082 on Build GRCh38
rs776734872 Chr.9: 137307212-137307212 on Build GRCh38
rs750065096 Chr.9: 137307260-137307260 on Build GRCh38
rs375289849 Chr.9: 137307160-137307160 on Build GRCh38
rs895756503 Chr.9: 137307158-137307158 on Build GRCh38
rs922539988 Chr.9: 137307106-137307106 on Build GRCh38
rs769199276 Chr.9: 137307176-137307176 on Build GRCh38
rs763809571 Chr.9: 137307217-137307217 on Build GRCh38
rs764644190 Chr.9: 137307227-137307227 on Build GRCh38
rs780838101 Chr.9: 137307251-137307251 on Build GRCh38
rs765336040 Chr.9: 137307217-137307217 on Build GRCh38
rs770529999 Chr.9: 137307101-137307101 on Build GRCh38
rs762882257 Chr.9: 137307167-137307167 on Build GRCh38
rs372089084 Chr.9: 137307250-137307250 on Build GRCh38
rs759616156 Chr.9: 137307213-137307213 on Build GRCh38
rs185546260 Chr.9: 137307222-137307222 on Build GRCh38
rs1054565277 Chr.9: 137307175-137307175 on Build GRCh38
rs377386003 Chr.9: 137307153-137307153 on Build GRCh38
rs771178108 Chr.9: 137307202-137307202 on Build GRCh38
rs865969248 Chr.9: 137307243-137307243 on Build GRCh38
rs745308228 Chr.9: 137307171-137307171 on Build GRCh38
rs150928549 Chr.9: 137307221-137307222 on Build GRCh38
rs779358702 Chr.9: 137307083-137307083 on Build GRCh38
rs758779381 Chr.9: 137307093-137307093 on Build GRCh38
rs766507998 Chr.9: 137307077-137307077 on Build GRCh38
rs780776396 Chr.9: 137307167-137307167 on Build GRCh38
rs768750030 Chr.9: 137307185-137307185 on Build GRCh38

View Full Product Details