Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179576633-179576888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL6A
Entrez Gene ID
86
Gene Name
actin like 6A
Gene Aliases
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K
UniGene
Hs.435326
Gene Chromosome Location
Chr.3: 179562880-179588405 on Build GRCh38
Transcripts
NM_004301.4 NM_177989.3 NM_178042.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3205856 c.733C>G p.Q245E Chr.3: 179576878-179576878 on Build GRCh38
4980397 c.740C>T p.T247M Chr.3: 179576885-179576885 on Build GRCh38
5026083 c.733C>T p.Q245* Chr.3: 179576878-179576878 on Build GRCh38
7317012 c.620G>A p.R207K Chr.3: 179576668-179576668 on Build GRCh38
1617229 c.629T>G p.F210C Chr.3: 179576677-179576677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775701995 Chr.3: 179576735-179576735 on Build GRCh38
rs373167296 Chr.3: 179576737-179576737 on Build GRCh38
rs879022012 Chr.3: 179576770-179576770 on Build GRCh38
rs916511838 Chr.3: 179576713-179576713 on Build GRCh38
rs34630517 Chr.3: 179576721-179576721 on Build GRCh38
rs878943667 Chr.3: 179576771-179576771 on Build GRCh38
rs755155418 Chr.3: 179576731-179576731 on Build GRCh38
rs370585441 Chr.3: 179576746-179576746 on Build GRCh38
rs745968297 Chr.3: 179576712-179576712 on Build GRCh38
rs746137886 Chr.3: 179576846-179576846 on Build GRCh38
rs768017323 Chr.3: 179576761-179576761 on Build GRCh38
rs778963657 Chr.3: 179576708-179576708 on Build GRCh38
rs375773279 Chr.3: 179576829-179576829 on Build GRCh38
rs997708202 Chr.3: 179576842-179576842 on Build GRCh38
rs751341025 Chr.3: 179576817-179576818 on Build GRCh38
rs772337037 Chr.3: 179576848-179576848 on Build GRCh38
rs1035631360 Chr.3: 179576824-179576824 on Build GRCh38
rs376110770 Chr.3: 179576738-179576738 on Build GRCh38
rs778205206 Chr.3: 179576813-179576813 on Build GRCh38
rs756664674 Chr.3: 179576805-179576805 on Build GRCh38
rs202173390 Chr.3: 179576791-179576791 on Build GRCh38
rs773187060 Chr.3: 179576742-179576742 on Build GRCh38
rs539611550 Chr.3: 179576690-179576690 on Build GRCh38
rs555771748 Chr.3: 179576794-179576794 on Build GRCh38
rs535487799 Chr.3: 179576781-179576781 on Build GRCh38
rs780324100 Chr.3: 179576862-179576862 on Build GRCh38
rs756597537 Chr.3: 179576676-179576676 on Build GRCh38
rs752198033 Chr.3: 179576795-179576795 on Build GRCh38
rs149451650 Chr.3: 179576700-179576700 on Build GRCh38
rs780193619 Chr.3: 179576759-179576760 on Build GRCh38
rs759098849 Chr.3: 179576793-179576793 on Build GRCh38
rs200004281 Chr.3: 179576707-179576707 on Build GRCh38
rs757498550 Chr.3: 179576833-179576833 on Build GRCh38
rs749632307 Chr.3: 179576697-179576697 on Build GRCh38
rs779212725 Chr.3: 179576838-179576838 on Build GRCh38
rs372804741 Chr.3: 179576782-179576782 on Build GRCh38

View Full Product Details