Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133414764-133415015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS13
Entrez Gene ID
11093
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 13
Gene Aliases
ADAM-TS13, ADAMTS-13, C9orf8, VWFCP, vWF-CP
UniGene
Hs.131433
Gene Chromosome Location
Chr.9: 133414339-133459403 on Build GRCh38
Transcripts
NR_024514.2 NM_001279349.1 NM_001279350.1 NM_001279351.1 NM_020385.3 NR_103995.1 NR_103996.1

Gene Symbol
REXO4
Entrez Gene ID
57109
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
REX4, XPMC2, XPMC2H
UniGene
Hs.438409
Gene Chromosome Location
Chr.9: 133406058-133418172 on Build GRCh38
Transcripts
NR_024514.2 NM_001279349.1 NM_001279350.1 NM_001279351.1 NM_020385.3 NR_103995.1 NR_103996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906859 c.454A>G p.T152A Chr.9: 133414783-133414783 on Build GRCh38
487188 c.386G>A p.R129K Chr.9: 133414851-133414851 on Build GRCh38
4163490 c.422G>A p.R141K Chr.9: 133414815-133414815 on Build GRCh38
5775896 c.305T>G p.I102S Chr.9: 133414932-133414932 on Build GRCh38
1701882 c.398C>T p.P133L Chr.9: 133414839-133414839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782783753 Chr.9: 133414840-133414840 on Build GRCh38
rs147890080 Chr.9: 133414904-133414904 on Build GRCh38
rs782403137 Chr.9: 133414993-133414993 on Build GRCh38
rs782532709 Chr.9: 133414895-133414895 on Build GRCh38
rs6597630 Chr.9: 133414815-133414815 on Build GRCh38
rs782514055 Chr.9: 133414903-133414903 on Build GRCh38
rs988432346 Chr.9: 133414812-133414812 on Build GRCh38
rs782206227 Chr.9: 133414977-133414978 on Build GRCh38
rs141547732 Chr.9: 133414905-133414905 on Build GRCh38
rs587596898 Chr.9: 133414828-133414828 on Build GRCh38
rs782612033 Chr.9: 133414819-133414819 on Build GRCh38
rs181933135 Chr.9: 133414878-133414878 on Build GRCh38
rs782094396 Chr.9: 133414853-133414853 on Build GRCh38
rs782263987 Chr.9: 133414811-133414811 on Build GRCh38
rs782516075 Chr.9: 133414980-133414980 on Build GRCh38
rs782439812 Chr.9: 133414929-133414929 on Build GRCh38
rs377306235 Chr.9: 133414800-133414800 on Build GRCh38
rs782287244 Chr.9: 133414928-133414928 on Build GRCh38
rs782202519 Chr.9: 133414968-133414968 on Build GRCh38
rs199989812 Chr.9: 133414845-133414845 on Build GRCh38
rs1027495353 Chr.9: 133414913-133414914 on Build GRCh38
rs782346333 Chr.9: 133414973-133414973 on Build GRCh38
rs782023241 Chr.9: 133414821-133414821 on Build GRCh38
rs782166709 Chr.9: 133414837-133414837 on Build GRCh38
rs147008170 Chr.9: 133414817-133414817 on Build GRCh38
rs374601411 Chr.9: 133414809-133414809 on Build GRCh38
rs781915967 Chr.9: 133414974-133414974 on Build GRCh38
rs782632484 Chr.9: 133414939-133414939 on Build GRCh38
rs781854599 Chr.9: 133414900-133414900 on Build GRCh38
rs370869145 Chr.9: 133414807-133414807 on Build GRCh38
rs782624296 Chr.9: 133414810-133414810 on Build GRCh38
rs782695524 Chr.9: 133414863-133414863 on Build GRCh38
rs782800841 Chr.9: 133414899-133414899 on Build GRCh38
rs782588604 Chr.9: 133414918-133414918 on Build GRCh38
rs909960522 Chr.9: 133414994-133414994 on Build GRCh38
rs781806472 Chr.9: 133414813-133414813 on Build GRCh38
rs782422263 Chr.9: 133414972-133414972 on Build GRCh38
rs1032708145 Chr.9: 133414791-133414791 on Build GRCh38
rs782431550 Chr.9: 133414801-133414801 on Build GRCh38
rs782443230 Chr.9: 133414917-133414917 on Build GRCh38

View Full Product Details