Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31687483-31687750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD16A
Entrez Gene ID
7920
Gene Name
abhydrolase domain containing 16A
Gene Aliases
BAT5, D6S82E, NG26, PP199
UniGene
Hs.388188
Gene Chromosome Location
Chr.6: 31686949-31703360 on Build GRCh38
Transcripts
NM_001177515.1 NM_021160.2 NR_033488.1 NR_033489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006357 c.1570C>T p.R524W Chr.6: 31687521-31687521 on Build GRCh38
4153254 c.1472T>G p.V491G Chr.6: 31687716-31687716 on Build GRCh38
6590856 c.1528G>A p.D510N Chr.6: 31687660-31687660 on Build GRCh38
7252375 c.1527C>T p.P509P Chr.6: 31687661-31687661 on Build GRCh38
7323402 c.1510C>T p.Q504* Chr.6: 31687678-31687678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955753379 Chr.6: 31687623-31687623 on Build GRCh38
rs754770819 Chr.6: 31687517-31687517 on Build GRCh38
rs375994936 Chr.6: 31687718-31687718 on Build GRCh38
rs775831073 Chr.6: 31687575-31687575 on Build GRCh38
rs763124323 Chr.6: 31687584-31687584 on Build GRCh38
rs143100378 Chr.6: 31687617-31687617 on Build GRCh38
rs777324878 Chr.6: 31687646-31687646 on Build GRCh38
rs62395815 Chr.6: 31687649-31687649 on Build GRCh38
rs763935494 Chr.6: 31687630-31687630 on Build GRCh38
rs576548642 Chr.6: 31687626-31687626 on Build GRCh38
rs372698997 Chr.6: 31687601-31687601 on Build GRCh38
rs544708496 Chr.6: 31687726-31687726 on Build GRCh38
rs10573 Chr.6: 31687661-31687661 on Build GRCh38
rs747546703 Chr.6: 31687534-31687534 on Build GRCh38
rs747264933 Chr.6: 31687680-31687680 on Build GRCh38
rs1033193502 Chr.6: 31687614-31687614 on Build GRCh38
rs146200640 Chr.6: 31687518-31687518 on Build GRCh38
rs56289010 Chr.6: 31687520-31687520 on Build GRCh38
rs746170043 Chr.6: 31687559-31687559 on Build GRCh38
rs972990257 Chr.6: 31687615-31687615 on Build GRCh38
rs970613978 Chr.6: 31687579-31687579 on Build GRCh38
rs764716626 Chr.6: 31687582-31687582 on Build GRCh38
rs774348828 Chr.6: 31687593-31687593 on Build GRCh38
rs775472010 Chr.6: 31687593-31687603 on Build GRCh38
rs1014885498 Chr.6: 31687557-31687557 on Build GRCh38
rs762851849 Chr.6: 31687615-31687616 on Build GRCh38
rs756797641 Chr.6: 31687663-31687663 on Build GRCh38
rs370156086 Chr.6: 31687619-31687619 on Build GRCh38
rs895817843 Chr.6: 31687555-31687555 on Build GRCh38
rs141653752 Chr.6: 31687655-31687655 on Build GRCh38
rs781751649 Chr.6: 31687667-31687667 on Build GRCh38
rs758057318 Chr.6: 31687645-31687645 on Build GRCh38
rs370817293 Chr.6: 31687560-31687560 on Build GRCh38
rs753802464 Chr.6: 31687616-31687616 on Build GRCh38
rs577318022 Chr.6: 31687674-31687674 on Build GRCh38
rs777292055 Chr.6: 31687536-31687536 on Build GRCh38
rs756957697 Chr.6: 31687725-31687725 on Build GRCh38
rs111601681 Chr.6: 31687546-31687546 on Build GRCh38
rs761962324 Chr.6: 31687596-31687596 on Build GRCh38
rs757906757 Chr.6: 31687521-31687521 on Build GRCh38
rs768777384 Chr.6: 31687591-31687591 on Build GRCh38
rs771400526 Chr.6: 31687535-31687535 on Build GRCh38
rs543642904 Chr.6: 31687644-31687644 on Build GRCh38
rs777193146 Chr.6: 31687531-31687531 on Build GRCh38

View Full Product Details