Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108862861-108863112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL7A
Entrez Gene ID
10881
Gene Name
actin like 7A
Gene Aliases
-
UniGene
Hs.123530
Gene Chromosome Location
Chr.9: 108862228-108863755 on Build GRCh38
Transcripts
NM_006687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7428105 c.657G>A p.V219V Chr.9: 108862979-108862979 on Build GRCh38
3902855 c.666C>T p.G222G Chr.9: 108862988-108862988 on Build GRCh38
752136 c.774C>G p.N258K Chr.9: 108863096-108863096 on Build GRCh38
278603 c.606G>A p.S202S Chr.9: 108862928-108862928 on Build GRCh38
5875304 c.676G>T p.V226L Chr.9: 108862998-108862998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777167099 Chr.9: 108863066-108863066 on Build GRCh38
rs762434806 Chr.9: 108863085-108863085 on Build GRCh38
rs761560935 Chr.9: 108862988-108862988 on Build GRCh38
rs995800010 Chr.9: 108863018-108863018 on Build GRCh38
rs749352487 Chr.9: 108862949-108862949 on Build GRCh38
rs201560933 Chr.9: 108863056-108863056 on Build GRCh38
rs773362387 Chr.9: 108862945-108862945 on Build GRCh38
rs772480894 Chr.9: 108862971-108862971 on Build GRCh38
rs1054744355 Chr.9: 108862922-108862922 on Build GRCh38
rs150012135 Chr.9: 108863013-108863013 on Build GRCh38
rs370929490 Chr.9: 108862891-108862891 on Build GRCh38
rs766265789 Chr.9: 108863006-108863006 on Build GRCh38
rs529306263 Chr.9: 108862914-108862914 on Build GRCh38
rs41278347 Chr.9: 108862962-108862962 on Build GRCh38
rs775752482 Chr.9: 108862913-108862913 on Build GRCh38
rs774892580 Chr.9: 108862899-108862899 on Build GRCh38
rs747366077 Chr.9: 108863063-108863063 on Build GRCh38
rs367596635 Chr.9: 108863040-108863040 on Build GRCh38
rs574614020 Chr.9: 108863039-108863039 on Build GRCh38
rs35639543 Chr.9: 108862961-108862962 on Build GRCh38
rs377392593 Chr.9: 108862928-108862928 on Build GRCh38
rs772745394 Chr.9: 108863084-108863084 on Build GRCh38
rs761482848 Chr.9: 108862927-108862927 on Build GRCh38
rs774093609 Chr.9: 108863091-108863091 on Build GRCh38
rs778033184 Chr.9: 108862948-108862948 on Build GRCh38
rs1009064437 Chr.9: 108863016-108863016 on Build GRCh38
rs759978598 Chr.9: 108862902-108862902 on Build GRCh38
rs112558776 Chr.9: 108862996-108862996 on Build GRCh38
rs767308676 Chr.9: 108863019-108863019 on Build GRCh38
rs371195181 Chr.9: 108862997-108862997 on Build GRCh38
rs757654506 Chr.9: 108862909-108862909 on Build GRCh38
rs201406481 Chr.9: 108862985-108862985 on Build GRCh38
rs766322447 Chr.9: 108863088-108863088 on Build GRCh38
rs750307981 Chr.9: 108862929-108862929 on Build GRCh38
rs751361858 Chr.9: 108863012-108863012 on Build GRCh38
rs753369277 Chr.9: 108862917-108862917 on Build GRCh38
rs747374066 Chr.9: 108862983-108862983 on Build GRCh38
rs757273596 Chr.9: 108862961-108862961 on Build GRCh38
rs372242791 Chr.9: 108863057-108863057 on Build GRCh38
rs763890352 Chr.9: 108862916-108862916 on Build GRCh38
rs769085021 Chr.9: 108863064-108863064 on Build GRCh38
rs3739693 Chr.9: 108862979-108862979 on Build GRCh38
rs776484063 Chr.9: 108862987-108862987 on Build GRCh38
rs747753886 Chr.9: 108863068-108863068 on Build GRCh38
rs766147732 Chr.9: 108862931-108862931 on Build GRCh38
rs932206963 Chr.9: 108862921-108862921 on Build GRCh38
rs746348823 Chr.9: 108862970-108862970 on Build GRCh38
rs561900425 Chr.9: 108862908-108862908 on Build GRCh38
rs529810713 Chr.9: 108862989-108862989 on Build GRCh38
rs779005184 Chr.9: 108863054-108863054 on Build GRCh38
rs200244439 Chr.9: 108862998-108862998 on Build GRCh38
rs145223364 Chr.9: 108863094-108863094 on Build GRCh38
rs753997762 Chr.9: 108863032-108863032 on Build GRCh38
rs566063853 Chr.9: 108862938-108862938 on Build GRCh38
rs547660889 Chr.9: 108862930-108862930 on Build GRCh38
rs371671871 Chr.9: 108863055-108863055 on Build GRCh38
rs769617223 Chr.9: 108863083-108863083 on Build GRCh38
rs756294901 Chr.9: 108862939-108862939 on Build GRCh38

View Full Product Details