Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1042320-1042577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA7
Entrez Gene ID
10347
Gene Name
ATP binding cassette subfamily A member 7
Gene Aliases
ABCA-SSN, ABCX, AD9
UniGene
Hs.134514
Gene Chromosome Location
Chr.19: 1040103-1065572 on Build GRCh38
Transcripts
NM_019112.3 XM_006722616.1 XM_006722617.2 XM_011527628.2 XM_011527629.1 XM_011527630.1 XM_011527631.1 XM_011527632.1 XM_011527633.2 XM_011527634.1 XM_011527635.1 XM_017026142.1 XM_017026143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990496 c.463G>A p.D155N Chr.19: 1042362-1042362 on Build GRCh38
69479 c.459G>A p.M153I Chr.19: 1042358-1042358 on Build GRCh38
1181268 c.485C>A p.S162* Chr.19: 1042384-1042384 on Build GRCh38
4388508 c.470C>T p.A157V Chr.19: 1042369-1042369 on Build GRCh38
3891439 c.483G>A p.T161T Chr.19: 1042382-1042382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538849668 Chr.19: 1042471-1042471 on Build GRCh38
rs775577517 Chr.19: 1042470-1042470 on Build GRCh38
rs996785033 Chr.19: 1042536-1042536 on Build GRCh38
rs151054304 Chr.19: 1042354-1042354 on Build GRCh38
rs765862595 Chr.19: 1042503-1042503 on Build GRCh38
rs780192436 Chr.19: 1042499-1042499 on Build GRCh38
rs775526158 Chr.19: 1042396-1042396 on Build GRCh38
rs779399216 Chr.19: 1042413-1042413 on Build GRCh38
rs769924680 Chr.19: 1042448-1042448 on Build GRCh38
rs772356677 Chr.19: 1042373-1042373 on Build GRCh38
rs73920555 Chr.19: 1042555-1042555 on Build GRCh38
rs756855704 Chr.19: 1042407-1042407 on Build GRCh38
rs938316552 Chr.19: 1042422-1042422 on Build GRCh38
rs776050442 Chr.19: 1042498-1042498 on Build GRCh38
rs758805228 Chr.19: 1042521-1042521 on Build GRCh38
rs1029706178 Chr.19: 1042537-1042537 on Build GRCh38
rs747299748 Chr.19: 1042434-1042434 on Build GRCh38
rs755444670 Chr.19: 1042362-1042362 on Build GRCh38
rs1026369358 Chr.19: 1042483-1042483 on Build GRCh38
rs759597670 Chr.19: 1042393-1042393 on Build GRCh38
rs922328283 Chr.19: 1042365-1042365 on Build GRCh38
rs758357947 Chr.19: 1042408-1042408 on Build GRCh38
rs140835505 Chr.19: 1042394-1042394 on Build GRCh38
rs755583115 Chr.19: 1042412-1042412 on Build GRCh38
rs369528566 Chr.19: 1042397-1042397 on Build GRCh38
rs751894726 Chr.19: 1042538-1042538 on Build GRCh38
rs758856011 Chr.19: 1042427-1042427 on Build GRCh38
rs964299399 Chr.19: 1042526-1042526 on Build GRCh38
rs1044005852 Chr.19: 1042367-1042367 on Build GRCh38
rs757598160 Chr.19: 1042540-1042540 on Build GRCh38
rs763834831 Chr.19: 1042341-1042341 on Build GRCh38
rs753273359 Chr.19: 1042520-1042520 on Build GRCh38
rs896492748 Chr.19: 1042458-1042458 on Build GRCh38
rs946881000 Chr.19: 1042392-1042392 on Build GRCh38
rs749872451 Chr.19: 1042358-1042358 on Build GRCh38
rs368818815 Chr.19: 1042401-1042401 on Build GRCh38
rs1056833797 Chr.19: 1042453-1042453 on Build GRCh38
rs781566880 Chr.19: 1042542-1042542 on Build GRCh38
rs776867096 Chr.19: 1042439-1042439 on Build GRCh38
rs778000491 Chr.19: 1042432-1042432 on Build GRCh38
rs553112069 Chr.19: 1042368-1042368 on Build GRCh38
rs973124893 Chr.19: 1042491-1042491 on Build GRCh38
rs747188678 Chr.19: 1042386-1042386 on Build GRCh38
rs375490467 Chr.19: 1042380-1042380 on Build GRCh38
rs776544869 Chr.19: 1042389-1042389 on Build GRCh38
rs751102411 Chr.19: 1042346-1042346 on Build GRCh38
rs750013923 Chr.19: 1042492-1042492 on Build GRCh38
rs763435591 Chr.19: 1042551-1042552 on Build GRCh38
rs896206697 Chr.19: 1042382-1042382 on Build GRCh38
rs771040546 Chr.19: 1042435-1042435 on Build GRCh38
rs778868953 Chr.19: 1042558-1042559 on Build GRCh38
rs770052706 Chr.19: 1042548-1042548 on Build GRCh38
rs4147905 Chr.19: 1042488-1042488 on Build GRCh38
rs780553526 Chr.19: 1042356-1042356 on Build GRCh38
rs140240982 Chr.19: 1042370-1042370 on Build GRCh38
rs764016385 Chr.19: 1042472-1042472 on Build GRCh38
rs778251252 Chr.19: 1042534-1042534 on Build GRCh38
rs1016805549 Chr.19: 1042506-1042506 on Build GRCh38
rs933695513 Chr.19: 1042366-1042366 on Build GRCh38
rs550075071 Chr.19: 1042543-1042543 on Build GRCh38
rs771112818 Chr.19: 1042388-1042388 on Build GRCh38
rs748597664 Chr.19: 1042418-1042418 on Build GRCh38
rs773985012 Chr.19: 1042487-1042487 on Build GRCh38

View Full Product Details