Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIGF
Entrez Gene ID
5281
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Gene Aliases
-
Chromosome Location
Chr.2: 46581274 - 46617112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002643.3 NP_002634.1 3 - 4 480 135
NM_173074.2 NP_775097.1 3 - 4 480 135
XM_005264369.2 XP_005264426.1 3 - 4 415 135
XM_011532908.2 XP_011531210.1 3 - 4 415 135
GenBank mRNA BC021725.1 - 3 - 4 135
BC029408.1 - 3 - 4 433 135
CR457000.1 - 2 - 3 316 135
D13435.1 - 3 - 4 383 135

View Full Product Details