The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173358.2 | NP_775494.1 | 4 - 5 | 439 | 127 |
XM_017029430.1 | XP_016884919.1 | 3 - 4 | 280 | 127 | |
XM_017029431.1 | XP_016884920.1 | 3 - 4 | 280 | 127 |